Skip to Content
Merck

Skip To

1003064

USP

Acetaminophen Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Nitrophenol, p-Nitrophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1281877
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor pressure

0.6 mmHg ( 120 °C)

API family

paracetamol, acetaminophen

autoignition temp.

541 °F

manufacturer/tradename

USP

bp

279 °C (lit.)

mp

110-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetaminophen Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR115035836241326
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

agriculture
environmental

application(s)

-

vapor pressure

0.6 mmHg ( 120 °C)

vapor pressure

0.6 mmHg ( 120 °C)

vapor pressure

0.6 mmHg ( 120 °C)

vapor pressure

0.6 mmHg ( 120 °C)

mp

110-115 °C (lit.)

mp

110-115 °C (lit.)

mp

110-115 °C (lit.)

mp

110-115 °C (lit.)


Still not finding the right product?

Explore all of our products under Acetaminophen Related Compound F


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 - STOT RE 2 Oral

target_organs

Kidney,Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

336.2 °F

flash_point_c

169 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1003064-50MG04061838662910

Questions

Reviews

No rating value

Active Filters