Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
vapor density
4.65 (vs air)
vapor pressure
1 mmHg ( 114 °C)
API family
acetaminophen, paracetamol
autoignition temp.
1004 °F
manufacturer/tradename
USP
bp
304 °C (lit.)
mp
113-115 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC(=O)Nc1ccccc1
InChI
1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)
InChI key
FZERHIULMFGESH-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| autoignition temp. 1004 °F | autoignition temp. 1004 °F | autoignition temp. 1004 °F | autoignition temp. 1004 °F |
| mp 113-115 °C (lit.) | mp 113-115 °C (lit.) | mp 113-115 °C (lit.) | mp ~Approximately 115 °C, 113-115 °C (lit.) |
Still not finding the right product?
Explore all of our products under Acetaminophen Related Compound D
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1003042-50MG | 04061838662903 |

