Skip to Content
Merck
All Photos(2)

Documents

241326

Sigma-Aldrich

4-Nitrophenol

ReagentPlus®, ≥99%

Synonym(s):

p-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein:
1281877
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39032018
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.6 mmHg ( 120 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder, crystals or chunks

autoignition temp.

541 °F

bp

279 °C (lit.)

mp

110-115 °C (lit.)

solubility

alcohol: freely soluble(lit.)
chloroform: freely soluble(lit.)
cold water: moderately soluble(lit.)
diethyl ether: freely soluble(lit.)

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

The degradation of 4-nitrophenol (4-NP) using cadmium sulphide nanoparticles was studied.

Application

4-Nitrophenol (4-NP) can be used to prepare 4-aminophenol (4-AP), a key intermediate for the manufacture of analgesic and antipyretic drugs.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

336.2 °F

Flash Point(C)

169 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

G Sangami et al.
Journal of nanoscience and nanotechnology, 14(3), 2299-2306 (2014-04-22)
An efficient method to degrade 4-nitrophenol (4-NP) using cadmium sulphide nanoparticles (CdS NPs) prepared by a novel method as a photocatalyst in the presence of H2O2 as a free radical generator was developed. To investigate the degradation mechanism, the interaction
Preparation of gold? dendrimer nanocomposites by laser irradiation and their catalytic reduction of 4-nitrophenol.
Hayakawa K, et al.
Langmuir, 19(13), 5517-5521 (2003)
Au/graphene hydrogel: synthesis, characterization and its use for catalytic reduction of 4-nitrophenol.
Li J, et al.
Journal of Materials Chemistry, 22(17), 8426-8430 (2012)
Reduction of 4-nitrophenol to 4-aminophenol over Au nanoparticles deposited on PMMA.
Kuroda K, et al.
J. Mol. Catal. A: Chem., 298(1-2), 7-11 (2009)
Kwok Wei Shah et al.
Materials (Basel, Switzerland), 12(10) (2019-05-28)
A novel synthesis of hexagonal gold nanoparticles (Au NPs) via hydrolyzed organosilane (3-mercaptopropyl)trimethoxysilane (MPTMS) using an ultrafast and environmentally friendly method is presented in this study. For the first time, organosilane MPTMS is used for chemical reduction of auric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service