Skip to Content
Merck

Skip To

1112467

USP

4-Chlorobenzophenone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(4-Chlorophenyl)phenylmethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
ClC6H4COC6H5
CAS Number:
Molecular Weight:
216.66
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
512043
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

cetirizine

manufacturer/tradename

USP

bp

195-196 °C/17 mmHg (lit.)

mp

74-76 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C13H9ClO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H

InChI key

UGVRJVHOJNYEHR-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

4-Chlorobenzophenone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C25007PHR26208.16116
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical small molecule

application(s)

-

format

neat

format

-

format

-

format

-

API family

cetirizine

API family

-

API family

cetirizine

API family

-

mp

74-76 °C (lit.)

mp

74-76 °C (lit.)

mp

74-76 °C (lit.)

mp

82-84 °C


Still not finding the right product?

Explore all of our products under 4-Chlorobenzophenone


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1112467-30MG04061833842331

Questions

Reviews

No rating value

Active Filters