Skip to Content
Merck

Skip To

1102952

USP

Cetirizine Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(2-{4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl}ethoxy)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H26ClN3O2
CAS Number:
Molecular Weight:
387.90
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

cetirizine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(N3CCN(CC3)CCOCC(=O)N)c2ccccc2

InChI

1S/C21H26ClN3O2/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)25-12-10-24(11-13-25)14-15-27-16-20(23)26/h1-9,21H,10-16H2,(H2,23,26)

InChI key

LVJDQBJDVOYDLA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetirizine Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Also, for use with USP monograph such as Cetirizine Hydrochloride Orally Disintegrating Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
110294111029631102930
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

API family

cetirizine

API family

cetirizine

API family

cetirizine

API family

cetirizine

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-


Still not finding the right product?

Explore all of our products under Cetirizine Related Compound C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1102952-25MG04061833842362

Questions

Reviews

No rating value

Active Filters