Skip to Content
Merck

Skip To

1102929

USP

Cetirizine dihydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

cetirizine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

room temp

SMILES string

OC(COCCN1CCN(C(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)CC1)=O.[H]Cl.[H]Cl

InChI

1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H

InChI key

PGLIUCLTXOYQMV-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetirizine dihydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cetirizine Hydrochloride
  • Levocetirizine Dihydrochloride

Biochem/physiol Actions

Cetirizine hydrochloride is a non-sedating type histamine H1-receptor antagonist.
Cetirizine hydrochloride is an orally active and selective H1-receptor antagonist. Antihistaminic; Piperazines. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C0980650PHR1656Y0001197
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

storage temp.

-

storage temp.

2-30°C

storage temp.

2-8°C

API family

cetirizine

API family

cetirizine

API family

cetirizine

API family

cetirizine


Still not finding the right product?

Explore all of our products under Cetirizine dihydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters