Skip to Content
Merck

Skip To

P110010

Polyvinyl butyral

Powder

Synonym(s):

PVB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
NA.47
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Polyvinyl butyral, resin prepared from vinyl acetate monomer

Quality Segment

form

powder

technique(s)

microbe id | staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

General description

Polyvinyl butyral is a type of resin that can be synthetically prepared from vinyl acetate monomer. It finds applications in the manufacture of phenolic resin bonded diamond wheels, used for grinding hard metals.

Application

Polyvinyl butyral resin similar to Butvar® B-98 for thin film support in electron microscopy.

Legal Information

Butvar is a registered trademark of Solutia, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M44451C5548O1875
technique(s)

microbe id | staining: suitable

technique(s)

titration: suitable

technique(s)

-

technique(s)

titration: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

form

powder

form

powder, crystals or chunks

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Plastic Films: Situation and Outlook : a Rapra Market Report (2004)
Phenolic Resins: Chemistry, Applications, Standardization, Safety and Ecology (2000)



Global Trade Item Number

SKUGTIN
T15601-500G04061837337857
821095000504022536473666
821095025004022536473673
821095999904022536473680
T15601-5G04061837841446
T15601-100G04061837337840
P110010-100G04061832343822

Questions

1–4 of 4 Questions  
  1. What is the solubility of the following chemicals: 1) Poly(9-vinylcarbazole) 2) P110010 -- Polyvinyl butyral?

    1 answer
    1. For Poly(9-vinylcarbazole) solubility, it is soluble in THF, chloroform, chlorobenzene, and dichlorobenzene. P110010, which is Polyvinyl butyral, is most soluble in a mixed solvent system such as ethanol/toluene, methanol/methyl ethyl ketone, etc.

      Helpful?

  2. Does Polyvinyl butyral bind to glass?

    1 answer
    1. Polyvinyl butyral is commonly used as a hot-melt adhesive to bind glass panels together for windows and other applications. It is known for its high tensile strength and its ability to bind glass splinters, making it a popular choice as an interlayer film in laminated safety glass.

      Helpful?

  3. what is the degree of acetylation and purity of the PVB? Density and molecular weight of the PVB?

    1 answer
    1. The only quality control testing performed for product P110010 is for appearance and viscosity. There is no testing performed on a lot-to-lot basis for density, purity, degree of acetylation, or molecular weight.
      The website does state that "Polyvinyl butyral resin similar to Butvar® B-98 for thin film support in electron microscopy."

      For Butvar B-98 the following information is available:
      Butvar® B-98 has the following characteristics (see https://productcatalog.eastman.com/tds/ProdDatasheet.aspx?product=71095422&pn=Butvar+-+B-98#_ga=2.208860571.827911750.1655887049-264082885.1655887049):
      - Molecular weight: 40,000 - 70,000 (Size exclusion chromatography with low angle laser light scattering standard)
      - % of hydroxyl content (as PV alcohol): 18.0 - 20.0%
      - % of acetate content (as PV acetate): 2.5% maximum

      Helpful?

  4. What is the molecular weight of thisPVA?

    1 answer
    1. Unfortunately, the molecular weight is not a measured characteristic for product P110010. However, as stated on the product detail page, "Polyvinyl butyral resin similar to Butvar® B-98 for thin film support in electron microscopy." B-98 has a reported molecular weight range of 40 kDa - 70 kDa.

      Please see the table on page 3 for reference: https://www.brenntag.com/media/documents/bsi/product_data_sheets/material_science/eastman/butvar_properties_and_uses.pdf

      Helpful?

Reviews

No rating value

Active Filters