Skip to Content
Merck
All Photos(2)

Key Documents

C5548

Sigma-Aldrich

Clobetasone butyrate

≥98% purity, powder

Synonym(s):

21-Chloro-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,11,20-trione 17-butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32ClFO5
CAS Number:
Molecular Weight:
478.98
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Clobetasone butyrate, ≥98%

Quality Level

Assay

≥98%

form

powder

color

white to faint yellow cast

mp

90-100 °C

solubility

chloroform: 50 mg/mL, clear, faintly yellow

ε (extinction coefficient)

13.4 at 243 nm in chloroform

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

InChI

1S/C26H32ClFO5/c1-5-6-22(32)33-26(21(31)14-27)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,28)20(30)13-24(19,26)4/h9-10,12,15,18-19H,5-8,11,13-14H2,1-4H3/t15-,18-,19-,23-,24-,25-,26-/m0/s1

InChI key

FBRAWBYQGRLCEK-AVVSTMBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clobetasone butyrate is a corticosteroid.

Application

Clobetasone butyrate has been used as an internal standard for the quantitative assay of clobetasol propionate.
Clobetasone butyrate has been used to analyze its bio-concentration factor (BCF) in Cyprinus carpio.

Biochem/physiol Actions

Clobetasone butyrate is a potent topical corticosteroid and is primarily used to treat atopic dermatitis (AD). Low dosage of this drug is used to treat dry eye in Sjogrens syndrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography--tandem mass spectrometric assay for clobetasol propionate in human serum from patients with atopic dermatitis
Sparidans RW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 878(23), 2150-2154 (2010)
Uptake and biological effects of synthetic glucocorticoids in common carp (Cyprinus carpio)
Nakayama K, et al.
Marine Pollution Bulletin, 85(2), 370-375 (2014)
Fluticasone propionate 0.05% cream once daily versus clobetasone butyrate 0.05% cream twice daily in children with atopic dermatitis
Wolkerstorfer A, et al.
Journal of the American Academy of Dermatology, 39(2), 226-231 (1998)
Safety and efficacy of 0.1% clobetasone butyrate eyedrops in the treatment of dry eye in Sjogren syndrome
Aragona P, et al.
European Journal of Ophthalmology, 23(3) (2013)
Community Pharmacy - E-Book: Symptoms Diagnosis and Treatment (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service