Skip to Content
Merck
All Photos(4)

Key Documents

O1875

Sigma-Aldrich

Orcinol monohydrate

≥99% purity (HPLC), powder

Synonym(s):

3,5-Dihydroxytoluene, 5-Methylresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,3-(OH)2 · H2O
CAS Number:
Molecular Weight:
142.15
Beilstein:
1071903
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Orcinol monohydrate, colorimetric detection reagent

biological source

synthetic

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

titration: suitable

color

white to light brown

bp

290 °C (lit.)

mp

56-58 °C (lit.)

solubility

H2O: 50 mg/mL

density

1.290 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.Cc1cc(O)cc(O)c1

InChI

1S/C7H8O2.H2O/c1-5-2-6(8)4-7(9)3-5;/h2-4,8-9H,1H3;1H2

InChI key

NBKPNAMTHBIMLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Orcinol is a small-molecule natural phenol involved in the production of a dye called orcein used for the staining of cells and chromosomes. Orcinol monohydrate is usually used to prepare a colorimetric reagent known as Bial′s reagent for the determination of RNA, DNA, and hexoses.

Application

Orcinol monohydrate has been used to prepare Bial′s reagent for Bial′s orcinol assay for pentose sugars. It is also used in the preparation of orcinol solution suitable for staining TLC plates and, measuring rhamnolipids in orcinol dye assays. Orcinol monohydrate has been used for the RNA assay.



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipopolysaccharide binding protein expression in primary human hepatocytes and HepG2 hepatoma cells.
Grube BJ
The Journal of Biological Chemistry, 269, 8477-8477 (1994)
Marjan De Mey et al.
Analytical biochemistry, 353(2), 198-203 (2006-03-21)
Recent developments in cellular and molecular biology require the accurate quantification of DNA and RNA in large numbers of samples at a sensitivity that enables determination on small quantities. In this study, five current methods for nucleic acid quantification were
Rapid colorimetric assays to qualitatively distinguish RNA and DNA in biomolecular samples.
Patterson Jennifer and Cameron Mura
Journal of Visualized Experiments (2013)
Estimation of monosaccharides by the orcinol-sulphuric acid reaction.
J BRUCKNER
The Biochemical journal, 60(2), 200-205 (1955-06-01)
R Almog et al.
Analytical biochemistry, 91(1), 130-137 (1978-11-01)
The standard orcinol test for estimating RNA is modified and developed as a specific method for the determination of RNA in the presence of DNA and proteins. The main differences in the procedure of the modified test when compared to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service