Skip to Content
Merck

Skip To

MSCAL4

ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids

MALDI MS Calibration Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ProteoMass

quality

MALDI MS Calibration Standard

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

General description

The ProteoMass MALDI Calibration Kit for Exactive Series, LTQ XL and LTQ hybrids enables calibration of Thermo Scientific ion trap and ion trap-based hybrid mass analyzers equipped with MALDI sources in positive and negative ion mode. It covers a mass range from m/z 144 to m/z 3,657, and is therefore usable for calibrations between m/z 50 and 4000.

Application

ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids has been used for tuning and calibration of LIT (linear ion trap) and Orbitrap mass spectrometer.[1][2][3]

Features and Benefits

Discover the Advantages for Yourself!
  • Developed and qualified specifically for use with Thermo Scientific Exactive Series MS, LTQ XL and LTQ hybrid mass spectrometers with MALDI sources
  • Contains high purity, low alkali metal solvents and recrystallized matrix
  • Conveniently packaged, to free you from time-consuming purification and weigh-up steps, and to allow you to focus on acquiring quality data

Legal Information

LTQ XL is a trademark of Thermo Fisher Scientific, Inc.
ProteoMass is a trademark of Sigma-Aldrich Co. LLC


Kit Components Only

Product No.
Description

  • Ethyl alcohol, Pure, 200 proof 10 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Acetonitrile 30 mL
    SDS

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Rachelle R Landgraf et al.
Analytical chemistry, 81(20), 8488-8495 (2009-09-16)
A hybrid linear ion trap/Orbitrap mass spectrometer was used to perform tandem mass spectrometry (MS/MS) experiments and high-resolution mass analysis of lipids desorbed from nerve tissue. A dramatic improvement in mass spectral resolution and a decrease in background are observed
Ruibing Chen et al.
Journal of proteome research, 9(2), 818-832 (2009-12-23)
The lobster Homarus americanus has long served as an important animal model for electrophysiological and behavioral studies. Using this model, we performed a comprehensive investigation of the neuropeptide expression and their localization in the nervous system, which provides useful insights
MALDI Produced Ions Inspected with a Linear Ion Trap-Orbitrap Hybrid Mass Analyzer
Strupat, K., et al.
Journal of the American Society For Mass Spectrometry, 20, 1451-1451 (2009)



Global Trade Item Number

SKUGTIN
MSCAL4-1KT04061826762301

Questions

Reviews

No rating value

Active Filters