Skip to Content
Merck
All Photos(1)

Key Documents

900666

Sigma-Aldrich

Bruker standard solvent

suitable for MALDI-MS, 50% ACN + 47.5% H2O + 2.5% TFA

Synonym(s):

Trifluoroacetic acid solution, TFA in H2O/ACN, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
5446934
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24
technique(s):
MALDI-MS: suitable
application(s):
general analytical
bp:
72.4 °C (lit.)
vapor pressure:
97.5 mmHg

vapor density

3.9 (vs air)

Quality Level

vapor pressure

97.5 mmHg

form

liquid

contains

2.5% trifluoroacetic acid
47.5% H2O
50% acetonitrile

technique(s)

MALDI-MS: suitable

bp

72.4 °C (lit.)

mp

-15 °C (lit.)

density

0.921 at 25 °C

cation traces

Ba: ≤5 ppm
Ca: ≤20 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤20 ppm
K: ≤20 ppm
Mg: ≤5 ppm
Mn: ≤5 ppm
Na: ≤20 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Zn: ≤5 ppm

application(s)

general analytical

SMILES string

OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoroacetic acid (TFA) is a nonaqueous solvent. At room temperature, TFA has the ability to liquify native and regenerated celluloses.

Biochem/physiol Actions

Trifluoroacetic acid (TFA) is mainly preferred as an internal chemical shift referencing agent in 19F NMR.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

19F-Site-Specific-Labeled Nucleotides for Nucleic Acid Structural Analysis by NMR
Test, 566, 59-87 (2016)
19F-Site-Specific-Labeled Nucleotides for Nucleic Acid Structural Analysis by NMR
Methods in Enzymology, 566, 59-87 (2016)
The use of biobased nanofibres in composites
Biofiber Reinforcements in Composite Materials, 571-647 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service