Skip to Content
Merck

Skip To

42597

3′-Deoxythymidine 5′-triphosphate sodium salt solution

BioChemika, puriss. p.a., suitable for MALDI-MS, ≥98.0% (HPLC)

Synonym(s):

2′,3′-Dideoxythymidine 5′-triphosphate, ddTTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H16N2NaO13P3
CAS Number:
Molecular Weight:
488.15
UNSPSC Code:
41116105
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioChemika

grade

puriss. p.a.

assay

≥98.0% (HPLC)

concentration

10.0-11.0 mmol/L (UV)

technique(s)

MALDI-MS: suitable

impurities

≤1.5% ddNDP (HPLC)

suitability

corresponds for UV test

storage temp.

−20°C

SMILES string

[Na+].CC1=CN([C@H]2CC[C@@H](COP([O-])(=O)OP(O)(=O)OP(O)(O)=O)O2)C(=O)NC1=O

InChI

1S/C10H17N2O13P3.Na/c1-6-4-12(10(14)11-9(6)13)8-3-2-7(23-8)5-22-27(18,19)25-28(20,21)24-26(15,16)17;/h4,7-8H,2-3,5H2,1H3,(H,18,19)(H,20,21)(H,11,13,14)(H2,15,16,17);/q;+1/p-1/t7-,8+;/m0./s1

InChI key

QJSZKDPCBDXBCK-KZYPOYLOSA-M

General description

3′-Deoxythymidine 5′-triphosphate sodium (ddTTP) is a sugar-modified dTTP analogue which is selective inhibitor of HIV-associated reverse transcriptase (HIV-RT).[1] It is also seen to inhibit the activity of DNA polymerase α.[2]

Packaging

packaging : 1 μmol (approx. 91-100 μL)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Elena N Ilina et al.
Frontiers in microbiology, 4, 186-186 (2013-07-13)
Spectinomycin remains a useful reserve option for therapy of gonorrhea. The emergence of multidrug-resistant Neisseria gonorrhoeae strains with decreased susceptibility to cefixime and to ceftriaxone makes it the only medicine still effective for treatment of gonorrhea infection in analogous cases.
Sylvia Steininger et al.
Nucleic acids research, 38(6), 1853-1865 (2009-12-31)
The Mre11/Rad50/Xrs2 (MRX) complex in Saccharomyces cerevisiae has well-characterized functions in DNA double-strand break processing, checkpoint activation, telomere length maintenance and meiosis. In this study, we demonstrate an involvement of the complex in the base excision repair (BER) pathway. We
Inhibition of the activity of DNA polymerase alpha by 2',3'-dideoxythymidine 5'-triphosphate.
K Ono et al.
Biochemical and biophysical research communications, 88(4), 1255-1262 (1979-06-27)



Global Trade Item Number

SKUGTIN
42597-1UMO-F04061833627129

Questions

Reviews

No rating value

Active Filters