Skip to Content
Merck

Skip To

E2758

β-Estradiol

BioReagent, powder, suitable for cell culture

Synonym(s):

1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
UNSPSC Code:
12352209
NACRES:
NA.77
PubChem Substance ID:
EC Number:
200-023-8
Beilstein/REAXYS Number:
1914275
MDL number:
Biological source:
synthetic (organic)
Form:
powder
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

product line

BioReagent

assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

mp

176-180 °C (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

O[C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(O)=CC=C4[C@@]3([H])CC[C@@]21C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

General description

β-Estradiol or estradiol-17 β (E2) is secreted majorly by the large preovulatory follicles in the ovary. In addition, estradiol can be synthesized de novo from cholesterol have androgens synthesized from cholesterol. The aromatization of androgen by enzyme aromatase (P450arom) results in the β-estradiol production.

Application

β-Estradiol is used to study cell differentiation and transformations (tumorigenicity).
β-Estradiol has been used:
  • as a component of Dulbecco′s modified Eagle medium (DMEM) for culturing mammary tumor cells
  • as a oestrogenic compound for testing neuroprotective effects
  • as a component of medium199 for culturing oocytes

Biochem/physiol Actions

The major estrogen secreted by the premenopausal ovary.
The major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation. Exposure to estradiol increases breast cancer incidence and proliferation.
β-Estradiol (E2) is a major premenopausal hormone in women and is essential for spermiogenesis, sperm maturation and motility in men. Cultured bone marrow stem cells (BMSCs) have ability to synthesize E2 and could be exploited for treating estrogen deficiency. It mediates physiological functions through estrogen receptor (ER) α and ERβ.

Physical form

powder-RT; stock-frozen in working aliquots, avoid repeated freeze/thaw

Preparation Note

To prepare a 20 μg/ml stock solution, add 1 ml absolute ethanol to 1 mg β-estradiol; gently swirl to dissolve; add 49 ml sterile medium while mixing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E2257E1132E1024
biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

-

biological source

-

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

product line

BioReagent

product line

BioXtra

product line

-

product line

-

form

powder

form

powder

form

-

form

powder

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-


Still not finding the right product?

Explore all of our products under β-Estradiol


pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
E2758-1G04061835568383
E2758-5G04061835514137
E2758-250MG04061835568390

Questions

1–5 of 5 Questions  
  1. Hello, you mention reconstitution in ug/ml. How much ethanol do I need to dissolve 250mg in to get a 100mM stock solution? Would I be able to do it directly in the glass bottle?

    1 answer
    1. To make a 100 mM stock solution with 250 mg of β-Estradiol (molecular weight 272.38 g/mol), dissolve it in 9.18 mL of ethanol. This is calculated as (0.250 g / 272.38 g/mol) / 0.1 mol/L = 0.00918 L or 9.18 mL. This can be done directly in a clean, dry glass bottle large enough to hold and mix the solution.

      Helpful?

  2. Can Estradiol/ethanol solutions be sterile filtered?

    1 answer
    1. Yes. stock solutions in ethanol may be sterile filtered. However, cellulose-based filter membranes are not recommended for use with ethanol. Please see the link below to review additional information regarding filter membrane compatibility with commonly used solvents:
      https://www.sigmaaldrich.com/technical-documents/technical-article/analytical-chemistry/filtration/chemical-compatibility-of-filter-components

      Helpful?

  3. Hello, I would like to check the solubility of this product in DMSO. Best,

    1 answer
    1. The solubility of this product in DMSO has not been determined. However, as per the literature, the β-Estradiol is soluble in DMSO at 20 mg/ml.

      Helpful?

  4. after dissolving the β-Estradiol in ethanol , whats the tempreture to store the item in ?

    1 answer
    1. The storage or stability conditions once the compound is in solution is not determined. General recommendations for β-Estradiol are to protect from light, and store at -20C or at 2-8C in a degassed solution. Solutions stored in this manner should remain stable in the range of 1 month - 1 year.

      Helpful?

  5. How diluit for concentration of the 100ng/mL

    1 answer
    1. β-Estradiol is soluble in absolute ethanol at 50 mg/mL (See E8875). Preparing a 100 ng/mL solution would require serial dilutions.
      Start with a 1 mg/mL (1000 ug/mL) stock in ethanol.
      Dilute 10 uL (10 ug = 10,000 ng) of this stock with 9.990 mL to achieve a 1000 ng/mL solution. This can be diluted with medium or buffer.
      Dilute 1 mL of this with 9 mL to the final concentration of 100 ng/mL.

      Helpful?

Reviews

No rating value

Active Filters