Skip to Content
Merck
All Photos(1)

Documents

E1132

Sigma-Aldrich

β-Estradiol

analytical standard

Synonym(s):

1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
Beilstein:
1914275
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.77

grade

analytical standard

packaging

vial of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

176-180 °C (lit.)

application(s)

pharmaceutical

SMILES string

O[C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(O)=CC=C4[C@@]3([H])CC[C@@]21C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation. Exposure to estradiol increases breast cancer incidence and proliferation.

Packaging

Supplied in amber screw-cap vials

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H R Andersen et al.
Environmental health perspectives, 107 Suppl 1, 89-108 (1999-05-07)
The aim of this study was to compare results obtained by eight different short-term assays of estrogenlike actions of chemicals conducted in 10 different laboratories in five countries. Twenty chemicals were selected to represent direct-acting estrogens, compounds with estrogenic metabolites
Yun Zhu et al.
PloS one, 12(2), e0171390-e0171390 (2017-02-06)
Mammalian lignans or enterolignans are metabolites of plant lignans, an important category of phytochemicals. Although they are known to be associated with estrogenic activity, cell signaling pathways leading to specific cell functions, and especially the differences among lignans, have not
Sarah B Putman et al.
PloS one, 10(10), e0140373-e0140373 (2015-10-16)
Because of poor reproduction after the lifting of an 8-year breeding moratorium, a biomedical survey of female lions in U.S. zoos was initiated in 2007. Fecal estrogen (FEM), progestagen (FPM) and glucocorticoid (FGM) metabolites were analyzed in samples collected 3-4
Nobutada Sakagami et al.
The Journal of veterinary medical science, 76(10), 1403-1405 (2014-07-01)
The influences of glucose supplementation on early development of bovine embryos in BSA-free synthetic oviduct fluid were examined. Among the groups supplemented with 1.5, 2.0, 4.0 or 5.6 mM glucose either at 0, 72 or 144 hr after fertilization, blastocysts
Pascal Pigeon et al.
Journal of medicinal chemistry, 48(8), 2814-2821 (2005-04-15)
A series of ruthenocene derivatives, 1-[4-(O(CH(2))(n)()N(CH(3))(2))phenyl]-1-(4-hydroxyphenyl)-2-ruthenocenylbut-1-ene, with n = 2-5, based on the structure of the breast cancer drug tamoxifen has been prepared. These compounds were obtained, via a McMurry cross-coupling reaction, as a mixture of Z and E isomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service