Skip to Content
Merck

Skip To

E1024

Estradiol

meets USP testing specifications

Synonym(s):

β-Estradiol, 1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
UNSPSC Code:
12352104
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-023-8
Beilstein/REAXYS Number:
1914275
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

USP/NF, meets USP testing specifications

form

powder

mp

176-180 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O[C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(O)=CC=C4[C@@]3([H])CC[C@@]21C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Biochem/physiol Actions

The major estrogen secreted by the premenopausal ovary.
The major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation. Exposure to estradiol increases breast cancer incidence and proliferation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E1132E1132E1132
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

-

form

-

form

-

mp

176-180 °C (lit.)

mp

176-180 °C (lit.)

mp

176-180 °C (lit.)

mp

176-180 °C (lit.)

agency

USP/NF, meets USP testing specifications

agency

-

agency

-

agency

-

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRB(2103), GPER(2852), SERPINA6(866)
mouse ... Esr1(13982), Esr2(13983), Esrra(26379)
rat ... Afp(24177), Ar(24208), Esr1(24890), Esr2(25149), Shbg(24775)

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRA(2101), ESRRB(2103), GPER(2852), SERPINA6(866)
mouse ... Esr1(13982), Esr2(13983), Esrra(26379)
rat ... Afp(24177), Ar(24208), Esr1(24890), Esr2(25149), Shbg(24775)

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRA(2101), ESRRB(2103), GPER(2852), SERPINA6(866)
mouse ... Esr1(13982), Esr2(13983), Esrra(26379)
rat ... Afp(24177), Ar(24208), Esr1(24890), Esr2(25149), Shbg(24775)

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRA(2101), ESRRB(2103), GPER(2852), SERPINA6(866)
mouse ... Esr1(13982), Esr2(13983), Esrra(26379)
rat ... Afp(24177), Ar(24208), Esr1(24890), Esr2(25149), Shbg(24775)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical


Still not finding the right product?

Explore all of our products under Estradiol


pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Fiona C Baker et al.
Psychoneuroendocrinology, 60, 96-104 (2015-07-05)
The menopausal transition is marked by increased prevalence in disturbed sleep and insomnia, present in 40-60% of women, but evidence for a physiological basis for their sleep complaints is lacking. We aimed to quantify sleep disturbance and the underlying contribution
Martial Pré et al.
Plant physiology, 147(3), 1347-1357 (2008-05-10)
Plant defense against pathogens depends on the action of several endogenously produced hormones, including jasmonic acid (JA) and ethylene. In certain defense responses, JA and ethylene signaling pathways synergize to activate a specific set of defense genes. Here, we describe
Hirotaka Ishii et al.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry, 13(4), 323-334 (2007-07-24)
It is believed that sex hormones are synthesized in the gonads and reach the brain via the blood circulation. In contrast with this view, the authors have demonstrated that sex hormones are also synthesized locally in the hippocampus and that



Global Trade Item Number

SKUGTIN
E1024-100G04061833601327
E1024-25G04061833601341
E1024-1G04061833601334

Questions

1–8 of 8 Questions  
  1. Is your estradiol and testosterone sterile?

    1 answer
    1. This product is not sterile. Prepared solutions would need to be filter-sterilized. Please see the link below to review a sterile Estradiol product option:
      https://www.sigmaaldrich.com/product/sigma/e2257

      Currently, a sterile testosterone product is not available. Testosterone product T1500, while not sterile or tested for use in cell culture, has been used successfully by other researchers in culture applications. Please see the link below to review an example publication:
      https://www.sciencedirect.com/science/article/pii/S0090955624033695?via%3Dihub

      Please see the link below to review this testosterone product option:
      https://www.sigmaaldrich.com/product/sigma/t1500

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Is product E1024 soluble in DMSO? If so, what is the recommended initial stock concentration to make?

    1 answer
    1. This product has not been tested for solubility in DMSO, however various sources report that it is soluble at 20 mg/mL.

      Helpful?

  5. What is the working range of Product E1024, Estradiol?

    1 answer
    1. For cell culture applications, the working range of this compound is 0.2 - 10 ng/mL.  For cell culture tested beta-estradiol, please refer to Sigma-Aldrich Prod. Nos. E2758 and E2257.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. What is the solution stability of Product E1024, Estradiol?

    1 answer
    1. Since this material is air sensitive, it is best to prepare solutions fresh.

      Helpful?

  8. What is Product E1024, Estradiol, soluble in?

    1 answer
    1. It is soluble in ethanol, acetone, dioxane, and sparingly in vegetable oils.

      Helpful?

Reviews

No rating value

Active Filters