Skip to Content
Merck

Skip To

C8982

α-Cyano-4-hydroxycinnamic acid

suitable for MALDI-TOF MS

Synonym(s):

α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HOC6H4CH=C(CN)CO2H
CAS Number:
Molecular Weight:
189.17
UNSPSC Code:
12352106
NACRES:
NA.21
PubChem Substance ID:
EC Number:
248-879-1
Beilstein/REAXYS Number:
3271427
MDL number:
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

analyte chemical class(es)

glycans, peptides, proteins

mp

245-250 °C (lit.)

suitability

suitable for MALDI-TOF MS

storage temp.

room temp

SMILES string

OC(=O)\C(=C\c1ccc(O)cc1)C#N

InChI

1S/C10H7NO3/c11-6-8(10(13)14)5-7-1-3-9(12)4-2-7/h1-5,12H,(H,13,14)/b8-5+

InChI key

AFVLVVWMAFSXCK-VMPITWQZSA-N

General description

α-Cyano-4-hydroxycinnamic acid (CHCA) is specially purified and qualified as a matrix for peptide MALDI-MS (matrix assisted laser desorption ionization-mass spectrometry) calibration standards. CHCA enhances ionization of protein or peptide standards and samples in matrix assisted laser desorption ionization (MALDI) mass spectrometers.[1]
It is also available as part of our ProteoMass MALDI calibration kits (MS-CAL1 and MS-CAL3).

Application

α-Cyano-4-hydroxycinnamic acid has been used as a matrix for MALDI-TOF MS (matrix assisted laser desorption ionization-time of flight mass spectrometry) analysis. The matrix has also been used for sample preparation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3946870990C2020
form

powder

form

solid

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

suitability

suitable for MALDI-TOF MS

suitability

-

suitability

-

suitability

-

analyte chemical class(es)

glycans, peptides, proteins

analyte chemical class(es)

glycans, peptides, proteins

analyte chemical class(es)

glycans, peptides, proteins

analyte chemical class(es)

-

mp

245-250 °C (lit.)

mp

245-250 °C

mp

245-250 °C

mp

245-250 °C (lit.)


Still not finding the right product?

Explore all of our products under α-Cyano-4-hydroxycinnamic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1B

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Utilizing isotope dilution-matrix-assisted laser desorption ionization-time of flight mass spectrometry as a reference procedure for the radioimmunoassay of serum thyroxine.
Tyan YC, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 420, 99-103 (2013)
a-Cyano-4-hydroxycinnamic acid as a matrix for matrixassisted laser desorption mass spectromtry.
Beavis RC, et al.
Journal of Mass Spectrometry : Jms, 27, 156-158 (1992)
MALDI-TOF MS sample preparation by using alkanethiolate self-assembled monolayers: A preliminary application for protein sample analysis.
Tyan Y-C, et al.
International Journal of Mass Spectrometry, 262, 67-72 (2007)



Global Trade Item Number

SKUGTIN
C8982-10X10MG04061835572359
C8982-10MG04061832011288

Questions

1–2 of 2 Questions  
  1. What can be used for solublization of α-Cyano-4-hydroxycinnamic acid?

    1 answer
    1. α-Cyano-4-hydroxycinnamic acid (α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA) is soluble in methanol (up to 50 mg/ml). It is also soluble at 10 mg/ml using 50% acetonitrile in 0.05% TFA for MALDI-MS. The acetonitrile concentration can be adjusted for individual preferences.For biological applications, it has been solubilized at 100 mM in DMSO or 50 mM in ethanol.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters