Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
vapor pressure
4.5 psi ( 20 °C)
Quality Level
assay
≥99%
form
liquid
contains
0.01% thymol as stabilizer
refractive index
n20/D 1.369 (lit.)
bp
50.2 °C (lit.)
density
1.872 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(Cl)Br
InChI
1S/C2HBrClF3/c3-1(4)2(5,6)7/h1H
InChI key
BCQZXOMGPXTTIC-UHFFFAOYSA-N
Gene Information
human ... ATP5D(513), GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879), GLRA1(2741), GLRB(2743), GRIA1(2890), KCNK10(54207), KCNK18(338567), KCNK2(3776), KCNK3(3777), KCNK9(51305), KCNN4(3783)
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99% | assay - | assay - | assay 97% |
| Quality Level 200 | Quality Level - | Quality Level - | Quality Level 100 |
| refractive index n20/D 1.369 (lit.) | refractive index - | refractive index - | refractive index n20/D 1.422 |
| bp 50.2 °C (lit.) | bp - | bp - | bp 75-80 °C/15 mmHg |
| density 1.872 g/mL at 25 °C (lit.) | density - | density - | density 1.031 g/mL at 20 °C (lit.), 1.024 g/mL at 25 °C |
| form liquid | form solid | form solid | form - |
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B4388-125ML | 04061833429631 |
| B4388-250ML | 04061833429648 |






