Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
levalbuterol, albuterol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Cl.CC(C)(C)NC[C@H](O)c1ccc(O)c(CO)c1
InChI
1S/C13H21NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,12,14-17H,7-8H2,1-3H3;1H/t12-;/m0./s1
InChI key
OWNWYCOLFIFTLK-YDALLXLXSA-N
Gene Information
human ... ADRB2(154)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Levalbuterol Inhalation Solution
Biochem/physiol Actions
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade - | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename USP |
| format neat | format - | format - | format neat |
| application(s) pharmaceutical (small molecule) | application(s) - | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) |
| Gene Information human ... ADRB2(154) | Gene Information human ... ADRB2(154) | Gene Information - | Gene Information human ... ADRB1(153), ADRB2(154), ADRB3(155) |
| API family levalbuterol, albuterol | API family - | API family - | API family isoproterenol |
Still not finding the right product?
Explore all of our products under Levalbuterol hydrochloride
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1358784-200MG | 04061833832295 |



