Skip to Content
Merck

Skip To

1012600

USP

Albuterol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Salbutamol, α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6405698
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

salbutamol, albuterol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

InChI

1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3

InChI key

NDAUXUAQIAJITI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Albuterol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

β2-adrenoceptor agonist

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S0100000BP451S5013
Salbutamol British Pharmacopoeia (BP) Reference Standard

BP451

Salbutamol

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

format

neat

format

neat

format

neat

format

-

Gene Information

human ... ADRB2(154)

Gene Information

human ... ADRB2(154)

Gene Information

human ... ADRB2(154)

Gene Information

human ... ADRB2(154)

API family

salbutamol, albuterol

API family

albuterol, salbutamol

API family

salbutamol, albuterol

API family

-


Still not finding the right product?

Explore all of our products under Albuterol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters