Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
salbutamol, albuterol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OS(O)(=O)=O.CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2
InChI
1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4)
InChI key
BNPSSFBOAGDEEL-UHFFFAOYSA-N
Gene Information
human ... ADRB2(154)
General description
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename EDQM | manufacturer/tradename BP |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| Gene Information human ... ADRB2(154) | Gene Information human ... ADRB2(154) | Gene Information human ... ADRB2(154) | Gene Information human ... ADRB2(154) |
| API family salbutamol, albuterol | API family albuterol, salbutamol | API family albuterol, salbutamol | API family albuterol, salbutamol |
signalword
Warning
hcodes
Hazard Classifications
Carc. 2 - Repr. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



