Skip to Content
Merck

Skip To

1133580

USP

Oversulfated Chondroitin Sulfate

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

heparin

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

General description

Oversulfated Chondroitin Sulfate (OSCS) is a prominent contaminant found in heparin that is connected to adverse clinical events. OSCS exhibits plasma anticoagulant activity and the capability to activate prekallikrein to kallikrein.

Oversulfated Chondroitin Sulfate USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia

Oversulfated Chondroitin Sulfate (OSCS) is a known contaminant in heparin products that led to safety concerns globally. This USP standard is used in identity and impurity testing to detect OSCS contamination using NMR or strong anion exchange chromatography.

The USP biologics carbohydrates category includes a variety of carbohydrate-based substances that are essential in the development and manufacturing of therapeutic products. These carbohydrates play crucial roles in biological processes and are often utilized as excipients, stabilizers, or active ingredients in pharmaceuticals. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of carbohydrate therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Oversulfated Chondroitin Sulfate USP reference standard suitable for use in specified USP compendial tests and assays. Also used to prepare system suitability solutions for the identification of heparin sodium by proton NMR (Nuclear magnetic resonance) spectroscopy and chromatography method.

Further information is available in the monograph, Heparin Sodium, USP43-NF38 - 2195 of the USP (United States Pharmacopeia).

Other Notes

Oversulfated Chondroitin Sulfate (OSCS) is a prominent contaminant found in heparin that is connected to adverse clinical events. OSCS exhibits plasma anticoagulant acti
Oversulfated Chondroitin Sulfate (OSCS) is a prominent contaminant found in heparin that is connected to adverse clinical events. OSCS exhibits plasma anticoagulant activity and the capability to activate prekallikrein to kallikrein. Oversulfated Chondroitin Sulfate USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia
Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
11335701304050Y0000280
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

EDQM

form

powder

form

-

form

solid

form

-

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

heparin

API family

chondroitin sulfate

API family

heparin

API family

chondroitin sulfate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Oversulfated Chondroitin Sulfate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(6), 2195-2195 (2018)
The effect of increasing the sulfation level of chondroitin sulfate on anticoagulant specific activity and activation of the kinin system
Hogwood J, et al.
Testing (2018)
The effect of increasing the sulfation level of chondroitin sulfate on anticoagulant specific activity and activation of the kinin system
Hogwood J, et al.
PLoS ONE (2018)



Global Trade Item Number

SKUGTIN
1133580-10MG04061833849491

Questions

Reviews

No rating value

Active Filters