Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
heparin
form
powder
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O
InChI key
JRTRSJGZMRQDHI-UHFFFAOYSA-N
General description
Specifically formulated for boron quantification in LMWHs, this reference standard supports specialized testing scenarios, such as trace element analysis in manufacturing processes or quality assurance settings. Accurate boron content monitoring ensures raw material purity and safety.
The USP biologics carbohydrates category includes a variety of carbohydrate-based substances that are essential in the development and manufacturing of therapeutic products. These carbohydrates play crucial roles in biological processes and are often utilized as excipients, stabilizers, or active ingredients in pharmaceuticals. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of carbohydrate therapeutics throughout their lifecycle.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| application(s) USP Biologics | application(s) USP Biologics | application(s) USP Biologics | application(s) USP Biologics |
| form powder | form powder | form powder | form powder |
| format neat | format neat | format neat | format neat |
| API family heparin | API family heparin | API family heparin | API family heparin |
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
