Skip to Content
Merck

Skip To

1304038

USP

Heparin sodium identification

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Heparin sodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

heparin

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O

InChI

1S/C26H42N2O37S5.Na/c1-4(30)27-7-9(31)13(6(56-23(7)39)3-55-67(43,44)45)58-26-19(65-70(52,53)54)12(34)16(20(62-26)22(37)38)60-24-8(28-66(40,41)42)15(63-68(46,47)48)14(5(2-29)57-24)59-25-18(64-69(49,50)51)11(33)10(32)17(61-25)21(35)36;/h5-20,23-26,28-29,31-

InChI key

JRTRSJGZMRQDHI-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Heparin Sodium is the sodium salt of sulfated glycosaminoglycans. It is a mixture of heterogeneous molecules varying in molecular weights with a combination of activities against different factors of the blood clotting cascade. It is present in mammalian tissues and is usually obtained from the intestinal mucosa or other suitable tissues of domestic mammals used for food by humans. Heparin sodium identification USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Heparin sodium identification is a reference standard used for structural and compositional verification of UFH. It enables spectroscopic and chromatographic analysis to ensure the identity of heparin in pharmaceutical products.

The USP biologics carbohydrates category includes a variety of carbohydrate-based substances that are essential in the development and manufacturing of therapeutic products. These carbohydrates play crucial roles in biological processes and are often utilized as excipients, stabilizers, or active ingredients in pharmaceuticals. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of carbohydrate therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Heparin sodium identification USP reference standard is used to prepare standard solution and system suitability solution for the identification and analysis of heparin sodium using specific methods as described in the United States Pharmacopeia (USP) monographs. Further information is available in chapter, Heparin Sodium, USP43-NF38 – 2195 of USP.

Other Notes

Heparin Sodium is the sodium salt of sulfated glycosaminoglycans. It is a mixture of heterogeneous molecules varying in molecular weights with a combination of activities against different factors of the blood clotting cascade. It is present in mammalian tissues and is usually obtained from the intestinal mucosa or other suitable tissues of domestic mammals used for food by humans. Heparin sodium identification USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Heparin Sodium is the sodium salt of sulfated glycosaminoglycans. It is a mixture of heterogeneous molecules varying in molecular weights with a combination of activities against different factors of the blood clotting cascade. It is present in mammalian
Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRE0027H02000001304107
description

United States Pharmacopeia (USP) Reference Standard

description

-

description

BRP, European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

diagnostic assay manufacturing

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

agency

USP

agency

-

agency

-

agency

USP

API family

heparin

API family

-

API family

heparin

API family

heparin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters