Skip to Content
Merck

Skip To

P1644

Percoll®

pH 8.5-9.5 (20 °C)

Synonym(s):

Percoll Density Gradient Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

aseptically filled

Quality Segment

form

colloidal ((Colloidal solution of silica coated with polyvinylpyrrolidone (PVP)) )

technique(s)

cell culture | mammalian: suitable

color

clear colorless to faint yellow

pH

8.5-9.5 (20 °C)

application(s)

hematology
histology

storage temp.

2-8°C

General description

Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. Percoll consists of colloidal silica particles of 15–30 nm diameter (23% w/w in water), which have been coated with polyvinylpyrrolidone (PVP). The PVP coating renders the product completely non-toxic and ideal for use with biological materials.

Application

Percoll® has been used in sperm preparation to study the energy requirements of bovine embryos.
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used in cell separation and organelle isolation.
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used to establish non-toxic low viscosity, low osmolarity density gradients that can be used to isolate cells, organelles and viruses. It has been used to separate inner and outer membrane vesicles from E. coli. Percoll is isosmotic throughout the gradient, non-toxic, and is easily removed from the purified materials.

Legal Information

Percoll is a registered trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4937P7828A8577
sterility

aseptically filled

sterility

aseptically filled

sterility

aseptically filled

sterility

aseptically filled

form

colloidal ((Colloidal solution of silica coated with polyvinylpyrrolidone (PVP)) )

form

colloidal

form

colloidal (Colloidal solution of silica coated with polyvinylpyrrolidone (PVP))

form

solution

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | plant: suitable

technique(s)

ELISA: suitable, electrophoresis: suitable, western blot: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

application(s)

hematology
histology

application(s)

-

application(s)

agriculture

application(s)

-


Still not finding the right product?

Explore all of our products under Percoll®


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



R Hovius et al.
Biochimica et biophysica acta, 1021(2), 217-226 (1990-01-29)
Rat liver mitochondria were isolated by a combination of differential and Percoll gradient centrifugation, resulting in a highly pure and intact preparation, as assessed by marker enzyme analysis, latency of cytochrome-c oxidase, respiratory control index and electron microscopy. Two different
Tanja Zöller et al.
International journal of molecular sciences, 19(3) (2018-03-02)
Microglia are the resident immune cells of the central nervous system (CNS) and participate in physiological and pathological processes. Their unique developmental nature suggests age-dependent structural and functional impairments that might contribute to neurodegenerative diseases. In the present study, we
T Hirano et al.
The Journal of cell biology, 106(4), 1171-1183 (1988-04-01)
A temperature-sensitive mutant nuc2-663 of the fission yeast Schizosaccharomyces pombe specifically blocks mitotic spindle elongation at restrictive temperature so that nuclei in arrested cells contain a short uniform spindle (approximately 3-micron long), which runs through a metaphase plate-like structure consisting



Global Trade Item Number

SKUGTIN
P1644-25ML04061838262059
P1644-500ML04061838262066
P1644-1L04061838262042
P1644-100ML04061838262035

Questions

1–10 of 11 Questions  
  1. What is the details about the manufacturing process of Percoll, specifically regarding the amount of PVP used during bead soaking or the approximate PVP concentration within Percoll?

    1 answer
    1. Percoll is produced by Cytiva, formerly known as GE Life Sciences. Details regarding the proprietary manufacturing process are not publicly available. However, it can be mentioned that the Percoll particles range in size from 15 to 30 nanometers. While the density of Percoll solution is measured, the specific information regarding the amount of coating on the silica particles is not provided.

      Helpful?

  2. What is procedure for sperm preparation for ivf with percoll gradient

    1 answer
    1. Please note that this product is intended for research use only. This material is not tested for the isolation of sperm samples. The following reference may be helpful:
      Chen. Y. et al., Int. J. Fertil., 37, 315-319 (1992).

      Please see the link below to review the product datasheet for additional information.
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/977/430/p1644pis.pdf

      Helpful?

  3. CAN I FREEZE PERCOLL (P1644)? I SEE IN OTHER REF THAT I CAN, BUT NOT IN THIS REF

    1 answer
    1. This material should not be stored frozen for long periods of time. As per the product information sheet, if stored at –20 °C, gradients form upon thawing, necessitating mixing the contents of the bottle before use. Please see the link below to review the product datasheet for further information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/426/291/percoll-product-info-sheet-ms.pdf

      Helpful?

  4. Percoll (P1644) 을 gradient 만드는데 사용했는데요, P4937도 같은 용도로 사용 가능한가요?

    1 answer
    1. Yes, product P4937 can be used for the same use as product P1644.

      Helpful?

  5. I often find crystals. Can I filter the solution before use? What filter size could I use without affecting the functionality of the product?

    1 answer
    1. Particles may be removed by filtration or low speed centrifugation. More information about the particles filtration can be found in the product data sheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/977/430/p1644pis.pdf
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/426/291/percoll-product-info-sheet-ms.pdf

      Helpful?

  6. When using Product P1644, Percoll®, what density should I use for my particular system?

    1 answer
    1. The product information sheet (under Documents, above) has a table of densities for different organelles and cell types.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. What is the difference between Percoll® Products P4937 and P1644?

    1 answer
    1. Product Nos. P1644 and P4937 are both Percoll. Product No. P4937 has additional testing for use in cell culture.

      Helpful?

  9. Can you help me calculate gradients for Percoll®, Product P1644?

    1 answer
    1. The product information sheet (under Documents, above) has an equation for calculating density of a given solution.

      Helpful?

  10. Is there a "best" salt solution to use with Product P1644, Percoll®?

    1 answer
    1. Percoll can be used with various salt formulations (PBS, 1.5 M NaCl, HBSS, etc.). Physiological saline and balanced salt solutions can be suggested.  Sucrose is included to prevent aggregation.  You can find detailed preparation instructions on the product information sheet (under Documents, above) for this product.

      Helpful?

1–10 of 11 Questions  

Reviews

No rating value

Active Filters