Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Percoll
®
Products
Product Category
density gradient media (4)
Brand
Sigma-Aldrich (6)
Color
Manufacturer
pH Value
Physical Form
Quality Segment
Application
Package Size
Sterilization Method
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
P4937
Percoll
®
,
pH 8.5-9.5 (25 °C), suitable for cell culture
Expand
View Pricing
P1644
Percoll
®
,
pH 8.5-9.5 (20 °C)
Expand
View Pricing
GE17-0891-01
Percoll
®
,
Cytiva 17-0891-01, pack of 1 L
Expand
View Pricing
GE17-0891-02
Percoll
®
,
Cytiva 17-0891-02, pack of 250 mL
Expand
View Pricing
P7828
Percoll
®
,
suitable for plant cell culture
Expand
View Pricing
GE17-0891-09
Percoll
®
,
Cytiva 17-0891-09, pack of 6 × 1 L
Expand
View Pricing