Product Name
Percoll®, suitable for plant cell culture
sterility
aseptically filled
form
colloidal (Colloidal solution of silica coated with polyvinylpyrrolidone (PVP))
technique(s)
cell culture | plant: suitable
pH
8.9 (20 °C)
application(s)
agriculture
storage temp.
2-8°C
Quality Level
Looking for similar products? Visit Product Comparison Guide
Application
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used in cell separation and organelle isolation.
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used to establish non-toxic low viscosity, low osmolarity density gradients that can be used to isolate cells, organelles and viruses. It has been used to separate inner and outer membrane vesicles from E. coli. Percoll is isosmotic throughout the gradient, non-toxic, and is easily removed from the purified materials.
Percoll is used in plant research for protoplast and organelle (chloroplast, mitochondria) separation.
Percoll® has been used in the density gradient centrifugation isolation of intact chloroplasts from green alga, decidual stromal cells and distinct populations of rooster sperm.
Legal Information
Percoll is a registered trademark of Cytiva
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Guangkun Yin et al.
Plant & cell physiology, 50(7), 1305-1318 (2009-06-13)
The development of mitochondria during seed germination is essential for plant growth. However, the developmental process is still poorly understood. Temperature plays a key role in soybean germination, and in this study we characterized the mitochondrial ultrastructure and proteome after
Xiang-Jie Yin et al.
Clinical proteomics, 18(1), 9-9 (2021-02-24)
Pregnancy is a complicated physiological process. The multifaceted regulation of maternal-fetal interface is of great importance for maintaining normal pregnancy and avoiding fetal rejection and secondary abortion. Previous studies have focused on the clinical features or pathological biomarkers of fetal
Sperm Collection of Differential Quality Using Density Gradient Centrifugation
Ahammad MU, et al.
Journal of Visualized Experiments, 145(141), e58833-e58833 (2018)
A rapid method for chloroplast isolation from the green alga Chlamydomonas reinhardtii
Mason CB, et al.
Nature Protocols, 1(5), 2227-2227 (2006)
The decidual stromal cells-secreted CCL2 induces and maintains decidual leukocytes into Th2 bias in human early pregnancy
He YY, et al.
Clinical Immunology (Orlando, Fla.), 145(2), 161-173 (2012)
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service