Skip to Content
Merck

Skip To

E5134

Ethylenediaminetetraacetic acid disodium salt dihydrate

suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)

Ethylenediaminetetraacetic acid disodium salt dihydrate

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2, Sequestrene Na2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
NACRES:
NA.31
PubChem Substance ID:
UNSPSC Code:
12352107
EC Number:
205-358-3
Beilstein/REAXYS Number:
3900609
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

assay

99.0-101.0% (titration)

form

powder

reaction suitability

reagent type: chelator

technique(s)

electrophoresis: suitable

impurities

≤0.005% Insolubles, ≤0.1% Nitrilotriacetic acid (NTA)

mp

248 °C (dec.) (lit.)

solubility

3  M NaOH: 1.6 g/10 mL

cation traces

Fe: ≤0.005%, Pb: ≤0.002%

suitability

suitable for electrophoresis, suitable for molecular biology

foreign activity

DNase, RNase, NICKase and protease, none detected

storage temp.

room temp

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

General description

EDTA disodium salt dehydrate chelates metal ions, forming an octahedral complex with divalent cations. This is useful in molecular biology research to inhibit enzymes dependent on activation by metal ions. In diagnostic assays, it acts as an anticoagulant during blood collection for hematological tests

Application

EDTA disodium salt has been used in research:
  • as a component in buffer solutions for electrophoresis, cell culture, enzymatic assays, siRNA, flow cytometry, dimetylmethylene blue, and more[1]
  • an anticoagulant for whole blood collection
  • a stabilizer solution for freezing cell culture samples[2]
  • in combination with PBS and trypsin, to trypsinize cells[3]
  • for phytoextraction of heavy metals in polluted soils
Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Features and Benefits

  • Molecular Biology grade, suitable for many molecular biology applications
  • Free of endonucleases DNase, RNase, Nickase, and Protease

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E6635E48844010-OP
grade

Molecular Biology

grade

Sigma Grade

grade

ACS reagent

grade

-

technique(s)

electrophoresis: suitable

technique(s)

cell culture | plant: suitable

technique(s)

-

technique(s)

-

assay

99.0-101.0% (titration)

assay

98.5-101.5%

assay

99.0-101.0%

assay

≥99.0%

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

100

solubility

3  M NaOH: 1.6 g/10 mL

solubility

H2O: soluble, clear, colorless

solubility

H2O: 100 g/L at 20 °C

solubility

-

impurities

≤0.005% Insolubles, ≤0.1% Nitrilotriacetic acid (NTA)

impurities

-

impurities

≤0.005% Insoluble matter, ≤0.1% Nitrilotriacetic acid ([HOCOCH2]3N)

impurities

-


Still not finding the right product?


pictograms

Exclamation markHealth hazard

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



External quality assessment of methylmalonic acid and total homocysteine
Clinical Chemistry, 45, 1536-1542 (1999)
Expression of ABCG2 and Bmi-1 in oral potentially malignant lesions and oral squamous cell carcinoma.
Dalley AJ, et al.
Cancer Medicine, 3(2), 273-283 (2014)
Ana Amaral et al.
Animals : an open access journal from MDPI, 11(1) (2021-01-21)
Neutrophils can originate neutrophil extracellular traps (NETs). Myeloperoxidase (MPO) is a peroxidase found in NETs associated to equine endometrosis and can be inhibited by 4-aminobenzoic acid hydrazide (ABAH). Metallopeptidases (MMPs) participate in extracellular matrix stability and fibrosis development. The objectives



Global Trade Item Number

SKUGTIN
E5134-5KG04061835544547
E5134-100G04061835544509
E5134-250G04061835544523
E5134-1KG04061835544516
E5134-50G04061835544530
E5134-500G04061838357540

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Product E5134 has a re-test date of Aug. 2023. Has the re-test been completed?

    1 answer
    1. The specific lot number is required to determine the retest status. You may obtain this lot status by contacting our Technical Service team. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information, including the specific lot number, so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

  4. What is the working concentration for Product E5134, EDTA disodium salt, for use as an anti-coagulant?

    1 answer
    1. The optimal concentration is 1.5 mg per mL of blood.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Is Product E5134, EDTA disodium salt, stable in solution?

    1 answer
    1. Yes. A 0.5 M stock solution at pH 8.5 is stable for months at 2-8°C.

      Helpful?

  7. Can solutions of Product E5134, EDTA disodium salt, be autoclaved?

    1 answer
    1. Yes.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review