Skip to Content
Merck
All Photos(1)

Documents

27285

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt dihydrate

meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
EC Number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77

Agency

tested according to BP
tested according to Ph. Eur.
tested according to USP

Quality Level

reg. compliance

tested according to FCC

Assay

99.0-101.0%

form

crystals

quality

meets analytical specification of Ph. Eur., BP, USP, FCC

reaction suitability

reagent type: chelator

impurities

residual solvents, complies
≤0.001% cyanide (CN)
≤0.001% heavy metals (as Pb)
≤0.1% nitrilotriacetic acid (HPLC) (HPLC)

loss

8.7-11.4% loss on drying, 200 °C, 3 h

pH

4-5 (20 °C, 5%)

mp

248 °C (dec.) (lit.)

solubility

water: soluble 100 g/L at 20 °C

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

As: ≤1 mg/kg
Ca:, in accordance
Fe: ≤10 mg/kg
Pb: ≤10 mg/kg

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid disodium salt dihydrate, referred to as EDTA is a widely used chelator of divalent cations like Ca⁺². It can inhibit proteases dependent on metal ions, such as, calcium-dependent cysteine proteases. EDTA also acts as a chelator of the Zn ion, inhibiting metalloproteases that require zinc for activity.

Application

Ethylenediaminetetraacetic acid disodium salt dihydrate has been used along with Na2HPO4 and citric acid for preparing Na2EDTA-McIlvaine buffer. Aqueous solution of Ethylenediaminetetraacetic acid disodium salt dihydrate has also been used for photocatalytic hydrogen production
Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haruthai Sangjarusvichai et al.
Talanta, 79(4), 1036-1041 (2009-07-21)
In this report, we aimed to extend our previous efforts toward the evaluation of sulfonamides (SAs) with a boron-doped diamond (BDD) electrode. We improved this method by reducing the analysis time using a monolithic column coupled with amperometric detection to
Solvent-free hydrothermal synthesis of anatase TiO2nanoparticleswith enhanced photocatalytic hydrogen production activity.
Ayad F. Alkaim
Applied Catalysis A: General, 466, 32- 37 (2013)
Sinan Akbayram et al.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis, 17(5), 494-496 (2010-06-10)
Ethylenediaminetetraacetic acid (EDTA)-dependent pseudothrombocytopenia (PTCP) is the phenomenon of a spurious low platelet count due to antiplatelet antibodies that cause platelet clumping in blood anticoagulated with EDTA. The aggregation of platelets in EDTA-dependent PTCP is usually prevented by other anticoagulants
Evaluation of EDTA as the chelator in the biuret reagent.
H Van Kley et al.
Clinical chemistry, 19(6), 621-623 (1973-06-01)
Suli Huang et al.
PloS one, 10(2), e0117007-e0117007 (2015-02-07)
microRNA (miRNA) plays a role in the pathogenesis of ischemic stroke, and single nucleotide polymorphisms in miRNA genes may contribute to disease susceptibility. However, the effect of miR-146a, miR-196a2, and miR-499 polymorphisms on ischemic stroke susceptibility has been rarely reported.

Articles

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service