Skip to Content
Merck

Skip To

E6635

Ethylenediaminetetraacetic acid disodium salt dihydrate

Sigma Grade, suitable for plant cell culture, 98.5-101.5%

Ethylenediaminetetraacetic acid disodium salt dihydrate
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352107
EC Number:
205-358-3
Beilstein/REAXYS Number:
3900609
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Sigma Grade

Quality Segment

assay

98.5-101.5%

form

powder

reaction suitability

reagent type: chelator

technique(s)

cell culture | plant: suitable

mp

248 °C (dec.) (lit.)

solubility

H2O: soluble, clear, colorless

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

General description

Ethylenediaminetetraacetic acid (EDTA) is an aminopolycarboxylic acid and a hexadentate ligand. It chelates with metal ions, especially with cations to form an octahedral complex. Ethylenediaminetetraacetic acid disodium salt (EDTA) is a blood anticoagulant and contributes to the pathogenesis of pseudothrombocytopenia. It chelates with calcium in the blood and inhibits clotting and is routinely used in haematological tests. It enhances the antibacterial activity of lysozyme. EDTA used in chelation therapy chelates with calcium and favors dilation of artery, solubilisation of atheromatous plaques in atherosclerotic vascular disease. EDTA chelation therapy may also protect from oxidative damage during blood and lipid peroxidation in liver fibrosis.

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Ethylenediaminetetraacetic acid disodium salt dihydrate has been in seed germination trials of plant species and in protein extraction from Moss, Physcomitrella paten. It has also been used in lysis and vacuole buffer for the isolation of vacuoles from Petunia petals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ED2SSE1644E4884
technique(s)

cell culture | plant: suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

Sigma Grade

grade

reagent grade

grade

-

grade

ACS reagent

assay

98.5-101.5%

assay

98.5-101.5% (titration)

assay

98.5-101.5%

assay

99.0-101.0%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mp

248 °C (dec.) (lit.)

mp

248 °C (dec.) (lit.)

mp

248 °C (dec.) (lit.)

mp

248 °C (dec.) (lit.)

solubility

H2O: soluble, clear, colorless

solubility

H2O: soluble, clear, colorless

solubility

H2O: 0.1 M, clear, colorless

solubility

H2O: 100 g/L at 20 °C


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Isolation of Intact Vacuoles from Petunia Petals and Extraction of Sequestered Glycosylated Phenylpropanoid Compounds
Skaliter O, et al.
Bio-protocol, 8(13), 56-56 (2017)
A Phosphopeptide Purification Protocol for the Moss Physcomitrella paten
Wang, X and He Y
Bio-protocol, 5(14), 56-56 (2015)
Induced Plant Accumulation of Lithium
Kavanagh L, et al.
Geostandards Newsletter, 8(2), 56-56 (2018)



Global Trade Item Number

SKUGTIN
E6635-1KG04061833604601
E6635-100G04061835544585
E6635-500G04061833604618

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review