Skip to Content
Merck

Skip To

DMN70

GenElute Direct mRNA Miniprep Kits

sufficient for 70 purifications

Synonym(s):

GenElute Direct mRNA Miniprep Kit, GenElute mRNA Kit, Gen Elute

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41105501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 70 purifications

Quality Segment

storage temp.

15-25°C

General description

Procedures such as cDNA synthesis, expression profiling and others require separation of mRNA from the vastly more abundant rRNA and tRNA. The GenElute mRNA kits provide convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA or directly from mammalian cells and tissues.

For direct mRNA preparation, cells or tissues are disrupted with SDS/proteinase K digestion to release RNA and eliminate RNases. The kit uses oligo (dT) covalently linked to 1 μm polystyrene beads to capture polyadenylated mRNA by hybridization. The polystyrene beads remain suspended during hybridization, eliminating the need for mixing or rocking, as is common for cellulose or magnetic particles. Polystyrene was also chosen because oligo(dT) polystyrene beads yield cleaner mRNA with fewer stringent washing steps than does the more commonly used oligo(dT) cellulose (2 or 3 wash steps versus 10 or more). With the GenElute kits, mRNA-bead complexes are washed on a microcentrifuge spin filter, and eluted into 10 mM Tris-HCL, pH 7.5.

Up to 107 mammalian cells or 40 mg tissue are lysed and homogenized, either with the filtration columns provided or with a mechanical homogenizer. RNase is eliminated during a 10 minute proteinase K digestion. Sodium chloride is added, and polyadenylated RNA is captured on oligo(dT) polystyrene beads during a 10 minute incubation. For further enrichment, RNA may be released from the beads into fresh lysis solution and recaptured with the original beads. After 3 washes in a spin column, purified mRNA is eluted in 100 μL of 10 mM Tris-HCl, pH 7.4.

The purified mRNA is ready for Northern analysis, reverse transcription and PCR, labeling for arrays, and other common applications.
The GenElute Direct mRNA Miniprep kit provides a convenient format to isolate polyadenylated mRNA directly from mammalian cells and tissues. The direct mRNA isolation procedure is based on that of Badley. Up to 107 mammalian cells or 40 mg tissue are lysed and homogenized, either with the filtration columns provided or with
a mechanical homogenizer. RNase is eliminated during a 10 minute proteinase K digestion. Sodium chloride is added, and polyadenylated RNA is captured on oligo(dT) polystyrene beads during a 10 minute incubation. For further enrichment, RNA may be released from the beads into fresh lysis solution and recaptured with the original beads. After 3 washes in a spin column, purified mRNA is eluted in 100 μl of 10 mM Tris-HCl, pH 7.4.

Application

GenElute Direct mRNA Miniprep Kit has been used to isolate mRNA from various tissues and cells.
The GenElute Direct Kit mRNA kit provides convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA or directly from mammalian cells and tissues.

Features and Benefits

  • Poly (A)+ mRNA isolated from total RNA in 40 minutes or 60 minutes directly from cells and tissues
  • Oligo(dT) polystyrene beads require fewer wash steps
  • mRNA captured on oligo(dT) polystyrene beads in 10 minutes, with no mixing or rocking
  • mRNA captured on oligo(dT) polystyrene beads in 10 minutes, with no mixing or rocking (Fig. 1)
  • Poly (A)+ mRNA isolated from total RNA in 40 minutes (Fig. 2) or 60 minutes directly from cells and tissues (Fig. 3)
  • Oligo(dT) polystyrene beads require fewer wash steps

Other Notes

For additional information, please see www.sigma-aldrich.com/mrna.
Use for isolating mRNA directly from mammalian cells or tissues.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MRN70DMN10MRN10
usage

sufficient for 70 purifications

usage

sufficient for 70 purifications

usage

sufficient for 10 purifications

usage

sufficient for 10 purifications

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

DMN10

MRN10

DMN70

MRN70


Still not finding the right product?


Kit Components Only

Product No.
Description

  • Collection tube 70 ea

  • Elution solution 10 mL

  • Filtration columns with tubes 70 ea

  • Lysis solution 120 mL

  • 5 M NaCl 8 mL

  • Oligo(dT)-polystyrene beads 2 mL

  • Proteinase K 25 mg

See All (11)

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
    SDS

  • Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
    SDS

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
DMN70-1KT04061833591604

Questions

Reviews

No rating value

Active Filters