Skip to Content
Merck
All Photos(1)

Key Documents

MRN70

Sigma-Aldrich

GenElute mRNA Miniprep Kit

sufficient for 70 purifications

Synonym(s):

GenElute mRNA Kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 70 purifications

technique(s)

RNA purification: suitable

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Procedures such as cDNA synthesis, expression profiling and others require separation of mRNA from the vastly more abundant rRNA and tRNA. The GenElute mRNA kits provide convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA or directly from mammalian cells and tissues. For direct mRNA preparation, cells or tissues are disrupted with SDS/proteinase K digestion to release RNA and eliminate RNases. Both kit types use oligo dT30 covalently linked to 1 μm polystyrene beads to capture polyadenylated mRNA by hybridization. The polystyrene beads remain suspended during hybridization, eliminating the need for mixing or rocking, as is common for cellulose or magnetic particles. Polystyrene was also chosen because oligo(dT) polystyrene beads yield cleaner mRNA with fewer stringent washing steps than does the more commonly used oligo(dT) cellulose (2 or 3 wash steps versus 10 or more). With the GenElute kits, mRNA-bead complexes are washed on a microcentrifuge spin filter, and eluted into 10 mM Tris-HCL, pH 7.5. mRNA prepared with either kit is suitable for a variety of downstream applications such as Northern blotting, expression array or chip hybridizations and cDNA synthesis and library construction.

Application

GenElute mRNA Miniprep Kit has been used to purify RNA from total RNA and to isolate RNA.
The purified mRNA is ready for Northern analysis, reverse transcription and PCR, labeling for arrays, and other common applications.

Features and Benefits

  • Quick and convenient reagent for use in the simultaneous isolation of RNA, DNA and protein
  • Performs well with large or small amounts of tissue or cells and many samples can be simultaneously extracted
  • One of the most effective methods for isolating total RNA. Purifications can be completed in only one hour starting with fresh tissue or cells

Other Notes

For additional information, please see www.sigma-aldrich.com/mrna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptome-wide mapping of N(6)-methyladenosine by m(6)A-seq based on immunocapturing and massively parallel sequencing.
Dominissini D
Nature Protocols, 8(1), 176-189 (2013)
K Oishi et al.
Journal of thrombosis and haemostasis : JTH, 4(7), 1566-1574 (2006-07-15)
Diabetes is associated with an excess risk of cardiac events, and one risk factor for infarction is an elevated level of plasminogen activator inhibitor-1 (PAI-1). To evaluate whether the glucocorticoid hormones are involved in the diabetes-induced PAI-1 production, we examined
Enjie Li et al.
eLife, 11 (2022-05-04)
Methyltransferase-like 3 (METTL3) and N6-methyladenosine (m6A) are involved in many types of biological and pathological processes, including DNA repair. However, the function and mechanism of METTL3 in DNA repair and chemotherapeutic response remain largely unknown. In present study, we identified
Adrenal gland-dependent augmentation of plasminogen activator inhibitor-1 expression in streptozotocin-induced diabetic mice.
Oishi K
Journal of Thrombosis and Haemostasis, 4(7), 1566-1574 (2006)
Miguel Angel Garcia-Campos et al.
Cell, 178(3), 731-747 (2019-07-02)
N6-methyladenosine (m6A) is the most abundant modification on mRNA and is implicated in critical roles in development, physiology, and disease. A major limitation has been the inability to quantify m6A stoichiometry and the lack of antibody-independent methodologies for interrogating m6A.

Articles

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service