Skip to Content
Merck
All Photos(1)

Documents

S5150

Sigma-Aldrich

Sodium chloride solution

5 M in H2O, BioReagent, for molecular biology, suitable for cell culture

Synonym(s):

Halite, NaCl, sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

liquid

concentration

5 M in H2O

technique(s)

ChIP: suitable
DNA extraction: suitable
cell culture | mammalian: suitable

impurities

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected
RNAse, none detected
<1.0 EU/mL Endotoxin level
<16 cfu/mL Bioburden

pH

6.0-8.0

density

1.190 g/cm3

anion traces

phosphate (PO43-): ≤5 ppm

cation traces

K: ≤50 ppm
heavy metals (as Pb): ≤5 ppm

suitability

suitable for Western blot
suitable for molecular biology

application(s)

agriculture
agriculture

foreign activity

DNAse, Exonuclease; RNAse; NICKase, Endonuclease; Protease, none detected

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium chloride solution has been used as a component of mycobacterial lysis buffer to extract DNA. It has also been used along with Dulbecco′s phosphate-buffered saline (DPBS) in the preparation of degassed buffer to elute plasma samples.

Preparation Note

Prepared in 18 megohm water.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gender-dependent effect of Gpbar1 genetic deletion on the metabolic profiles of diet-induced obese mice
Vassileva G, et al.
The Journal of Endocrinology, 205(3), 225-232 (2010)
Tim J Bull et al.
Journal of clinical microbiology, 41(7), 2915-2923 (2003-07-05)
Mycobacterium avium subsp. paratuberculosis is a robust and phenotypically versatile pathogen which causes chronic inflammation of the intestine in many species, including primates. M. avium subsp. paratuberculosis infection is widespread in domestic livestock and is present in retail pasteurized cows'
Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Laurent Dortet et al.
Journal of clinical microbiology, 52(7), 2359-2364 (2014-04-25)
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25

Protocols

Procedure and protocol for Anti Ago-RNA Immunoprecipitation from mammalian cells using the RIP kit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service