Skip to Content
Merck

Skip To

80948

Sodium sulfate

suitable for HPLC, LiChropur, 99.0-101.0% (T)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
UNSPSC Code:
12352302
NACRES:
NA.21
EC Number:
231-820-9
MDL number:
Assay:
99.0-101.0% (T)
Grade:
reagent grade
Form:
solid
Solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Level

assay

99.0-101.0% (T)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

mp

884 °C (lit.)

solubility

H2O: 1 M, clear, colorless

density

2.68 g/mL at 25 °C (lit.)

UV absorption

λ: 220 nm Amax: ≤0.1, λ: 230 nm Amax: ≤0.05, λ: 240 nm Amax: ≤0.03, λ: 250 nm Amax: ≤0.02, λ: 500 nm Amax: ≤0.02

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S56401346413464-M
form

solid

form

powder

form

crystalline

form

crystalline

solubility

H2O: 1 M, clear, colorless

solubility

water: soluble 1 g/10 mL

solubility

-

solubility

-

assay

99.0-101.0% (T)

assay

≥99.0%

assay

99.0-100.5% (calc. to the dried substance)

assay

99.0-100.5% (calc. to the dried substance)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

grade

reagent grade

grade

-

grade

-

grade

-

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)


Still not finding the right product?

Explore all of our products under Sodium sulfate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
85548-250ML04061833025567
840123100004022536710570
09569-50ML04061838210470
85548-1L04061833025512
S6760-1L04061837609398
S6760-250ML04061837609404
840123010004022536625164
840123999904022536708782
840123050004022536748504
840123000004022536625157

Questions

Reviews

No rating value

Active Filters