If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
liquid
Quality Segment
shelf life
limited shelf life, expiry date on the label
concentration
0.98-1.02 M
technique(s)
HPLC: suitable
color
colorless
pH
6.9-7.1
solubility
water: soluble at 60 °C
density
1.002 g/cm3 at 20 °C
λ
neat
UV absorption
λ: 260 nm Amax: ≤0.025, λ: 280 nm Amax: ≤0.030
storage temp.
2-8°C
General description
Application
- In reversed-phase chromatography for the analysis of nucleotide sugars.
- In the analysis of oligonucleotides using ultrahigh-performance liquid chromatography (UHPLC).
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) HPLC: suitable | technique(s) electrophoresis: suitable | technique(s) HPLC: suitable | technique(s) - |
| concentration 0.98-1.02 M | concentration 1 M, ~1.0 M in H2O | concentration 0.95-1.05 M | concentration ~1.0 M in H2O |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life - | shelf life - |
Still not finding the right product?
Explore all of our products under Triethylammonium acetate buffer
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-



