Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium sulfate
Linear Formula:
Na
2
SO
4
CAS Number:
7757-82-6
Molecular Weight:
142.04
EC Number:
231-820-9
Products
Product Category
salts (18)
buffers (1)
Brand
SAFC (1)
Sigma-Aldrich (14)
Supelco (4)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Anhydrous (10)
ACS reagent (4)
Puriss (2)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
239313
Sodium sulfate,
ACS reagent, ≥99.0%, anhydrous, granular
Expand
View Pricing
238597
Sodium sulfate,
ACS reagent, ≥99.0%, anhydrous, powder
Expand
View Pricing
1.06649
Sodium sulfate,
anhydrous for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
S9627
Sodium sulfate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
13464
Sodium sulfate,
puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Expand
View Pricing
746363
Sodium sulfate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
1.06637
Sodium sulfate,
anhydrous, coarse granules for analysis EMSURE
®
ACS
Expand
View Pricing
793531
Sodium sulfate,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
204447
Sodium sulfate,
≥99.99% trace metals basis
Expand
View Pricing
1.06645
Sodium sulfate,
anhydrous, fine powder, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JPE, USP
Expand
View Pricing
71959
Sodium sulfate,
BioUltra, anhydrous, ≥99.0% (T)
Expand
View Pricing
1.06639
Sodium sulfate,
anhydrous granulated for organic trace analysis EMSURE
®
Expand
View Pricing
8.22286
Sodium sulfate,
anhydrous for synthesis
Expand
View Pricing
S6547
Sodium sulfate,
BioXtra, ≥99.0%
Expand
View Pricing
1.06647
Sodium sulfate,
anhydrous 99.99 Suprapur
®
Expand
View Pricing
798592
Sodium sulfate,
anhydrous, granular, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
80948
Sodium sulfate,
suitable for HPLC, LiChropur
™
, 99.0-101.0% (T)
Expand
View Pricing
S5640
Sodium sulfate,
≥99.0%, suitable for plant cell culture
Expand
View Pricing
71963
Sodium sulfate,
tested according to Ph. Eur., anhydrous
Expand
View Pricing
13464-M
Sodium sulfate,
puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Expand
View Pricing