Skip to Content
Merck

Skip To

D6421

DMEM/F-12

With HEPES, sodium bicarbonate and sodium pyruvate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12, DME/F-12, 1:1 mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

sodium pyruvate: 0.055 g/L
L-glutamine: no
phenol red: yes
NaHCO3: yes
HEPES: 15 mM
glucose: 3.15 g/L

shipped in

ambient

storage temp.

2-8°C

General description

A measured mixture of chemically defined ingredients makes up a basal medium, which is a type of complex media. DMEM is a medium supplemented with serum and sometimes with tryptose phosphate broth (TPB). DMEM/F-12 is a completely defined synthetic medium, which is protein-free.[1]

Application

DMEM:F12 is a 50:50 mixture of DMEM and Ham′s F12 media that has proven to be useful in a wide range of cell culture applications, especially when supplemented with fetal bovine serum (FBS). Ham′s Nutrient Mixture F12 was originally developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells, and mouse L cells. It is frequently used with dialyzed serum, hormones, selenium, and other supplements for serum-free cultures. It is the medium of choice for supporting the growth of cells of rodent origin, particularly rabbit and rat, and has been proven to be an excellent cloning medium for myeloma and hybridoma cells.
Suitable for growth of wide range of cell types in low serum conditions.
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham is used in cell culture & proliferation assays.[2][3][4][5] It is also used in bone marrow-derived macrophages culture.[6]

Preparation Note

Supplement with 0.365 g/L L-glutamine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D8062D8437D8900
sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

powder

components

sodium pyruvate: 0.055 g/L
L-glutamine: no
phenol red: yes
NaHCO3: yes
HEPES: 15 mM
glucose: 3.15 g/L

components

glucose: 3.15 g/L
NaHCO3: yes
L-glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: 0.055 g/L

components

glucose: 3.15 g/L
HEPES: yes
L-glutamine: yes
phenol red: yes
sodium pyruvate: 0.055 g/L

components

sodium pyruvate: 0.055 g/L
HEPES: 15 mM
NaHCO3: no
L-glutamine: yes
glucose: 3.15 g/L
phenol red: yes

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yohei Ueda et al.
Scientific reports, 9(1), 8547-8547 (2019-06-14)
Growth retardation is an important side effect of glucocorticoid (GC)-based drugs, which are widely used in various preparations to treat many pediatric diseases. We investigated the therapeutic effect of exogenous CNP-53, a stable molecular form of intrinsic CNP, on a
E Zweygarth et al.
The Onderstepoort journal of veterinary research, 70(2), 165-168 (2003-09-12)
The Welgevonden stock of Ehrlichia ruminantium, aetiological agent of heartwater, was propagated in baby hamster kidney (BHK) cells, Chinese hamster ovary (CHO-K1) cells and Madin Darby bovine kidney (MDBK) cells. The cultures required supplementation of the medium with cycloheximide for
Kouichi Hasegawa et al.
Stem cells translational medicine, 1(1), 18-28 (2012-12-01)
An optimal culture system for human pluripotent stem cells should be fully defined and free of animal components. To date, most xeno-free culture systems require human feeder cells and/or highly complicated culture media that contain activators of the fibroblast growth



Global Trade Item Number

SKUGTIN
D6421-500ML04061833587522
D6421-24X500ML04061833587515
D6421-6X500ML04061833587539

Questions

1–2 of 2 Questions  
  1. Is it possible to get a complete list of media components, such as the amino acids and their concentrations please. I could not find it in the specification sheet.

    1 answer
    1. Please see the link below to review the Product Information sheet which lists the complete formulation, including amino acids and the associated concentrations:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/171/436/d6421for.pdf

      Helpful?

  2. May I confirm if it is acceptable to use 0.1 M HCl and 0.1 M NaOH to balance the pH of the medium (D6421)? Are there any specific recommendations or precautions for this adjustment process to maintain the integrity and performance of the medium?

    1 answer
    1. The pH of DMEM F-12 can be adjusted to 6.9 - 7.1 using NaOH 1N or HCl 1N.
      The pH of bicarbonate buffered solutions usually rises 0.1 - 0.2 units during filtration
      It is recommended to add HCl or NaOH drop-wise while continuously stirring to avoid drastic pH swings. Use the smallest volume possible to avoid significantly altering the osmolarity of the medium. Maintain sterile conditions when adjusting pH for cell culture use.

      Helpful?

Reviews

No rating value

Active Filters