Skip to Content
Merck

Skip To

D8437

DMEM/F-12

With ʟ-glutamine, HEPES, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12, DME/F-12, 1:1 mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: 3.15 g/L
HEPES: yes
L-glutamine: yes
phenol red: yes
sodium pyruvate: 0.055 g/L

shipped in

ambient

storage temp.

2-8°C

General description

Dulbecco′s Modified Eagle′s medium (DMEM), a regularly used cell culture medium, is a source of energy-rich media that includes glucose, vitamins, and amino acids.[1] A 1:1 mixture of Dulbecco′s Modified Eagle′s medium (DME) and Ham′s F-12 nutrient mixture is commonly used in cell culture research. To compensate for the loss of buffering capacity caused by removing serum, 4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES) buffer may be added in the formulations at a final concentration of 15 mM.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham:
  • has been used as a component in the culture medium to culture primary culture-activated human hepatic stellate cells[1]
  • has been used to culture the cartilage biopsies[2]
  • may be used for the propagation of Cowdria ruminantium[3]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D8062D8900D6434
sterility

sterile-filtered

sterility

sterile-filtered

sterility

-

sterility

sterile-filtered

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

powder

form

liquid

components

glucose: 3.15 g/L
HEPES: yes
L-glutamine: yes
phenol red: yes
sodium pyruvate: 0.055 g/L

components

glucose: 3.15 g/L
NaHCO3: yes
L-glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: 0.055 g/L

components

sodium pyruvate: 0.055 g/L
HEPES: 15 mM
NaHCO3: no
L-glutamine: yes
glucose: 3.15 g/L
phenol red: yes

components

L-glutamine: no
phenol red: no
NaHCO3: yes
HEPES: 15 mM
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–7 of 7 Questions  
  1. Dear, Can you tell me the composition of this medium, or at least the amount of L-Glutamine present in this media? Kind regards, Sarah

    1 answer
    1. Helpful?

  2. Is the glutamine that has already been added stable glutamine? How long is the shelf life?

    1 answer
    1. This product does not contain Alanyl-glutamine, which is considered a more stable form It has been supplemented with L-glutamine and will remain stable through out the shelf life of the product with proper storage and handling. Typical lots of D8437 are assigned an expiration date of 1 year from the date of release. Note that under culture conditions of 37 deg C the L-glutamine will gradually degrade. See the link below to review general stability information regarding L-glutamine.:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/195/924/t075.pdf

      Alanyl-glutamine is also available as a sterile medium supplement. Please see the link below for an example of one cell culture product option:
      https://www.sigmaaldrich.com/product/sigma/g8541

      Helpful?

  3. Can Product D8437, Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, be used under serum free conditions?

    1 answer
    1. When DME/F12 is used under serum free conditions, supplementation is necessary. DME/F12 can be used for growing the epithelial cell line TM4 (mouse testes) when supplemented with insulin, transferrin, EGF, FSH, somatomedin C, ovine growth hormone, and retinoic acid. Primary cultures of Sertoli cells can also be grown in the above supplemented media. Rat glial C6 cells have been grown in DME/F12 supplemented with transferrin, insulin, LH, and rat submaxillary gland extract. See: Jakoby, W.B., and Pastan, I.H., Cell Culture, Methods in Enzymology, 58, 103-106 (1979). (This book is listed as Product No. C0541).

      Helpful?

  4. What is the storage temperature of Product D8437, Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham?

    1 answer
    1. We recommend storing Product No. D8437 at 2-8°C.

      Helpful?

  5. What is Product D8437, Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham?

    1 answer
    1. This product is a 1:1 ratio mixture of Dulbecco’s Modified Eagle’s Medium (DME) high glucose and Nutrient Mixture F-12 Ham (F-12).

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. What is the optimal carbon dioxide concentration needed for Product D8437, Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham?

    1 answer
    1. This is a 1:1 mixture of the DME and F-12. The optimal carbon dioxide required for DME is 10% and for F-12 is 5%. Since this medium is a mixture, the optimal carbon dioxide concentration is typically 5% to 8%.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review