Skip to Content
Merck
All Photos(1)

Key Documents

D8062

Sigma-Aldrich

DMEM/F-12

With ʟ-glutamine, sodium bicarbonate and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: 3.15 g/L
NaHCO3: yes
L-glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: 0.055 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Media are supplemented with a defined mixture of nutrients, growth factors, and hormones instead of serum. A 1:1 mixture of Dulbecco′s Modified Eagle′s medium (DME) and Ham′s F-12 nutrient mixture is commonly used for these defined studies. To compensate for the loss of buffering capacity caused by eliminating serum, a final concentration of 15 mM HEPES buffer can be added to the formulations. It allows the investigator to optimize the buffering system.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham has been used:
  • to maintain human lung and colorectal cancer cell lines for the biological evaluation of the secondary metabolites
  • to culture human kidney-2 (HK-2) cells and as a component of the assay medium for lysophosphatidylcholine (LPC) treatment
  • to culture mouse squamous cell carcinoma (SCCVII) tumor cells

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new macrolide isolated from the endophytic fungus Colletotrichum sp
Bungihan ME, et al.
3rd Kuala Lumpur International Conference on Biomedical Engineering 2006: Biomed 2006, 6(1), 57-73 (2013)
Hayato Fukusumi et al.
PeerJ, 6, e4187-e4187 (2018-01-10)
Since the development of human-induced pluripotent stem cells (hiPSCs), various types of hiPSC-derived cells have been established for regenerative medicine and drug development. Neural stem/progenitor cells (NSPCs) derived from hiPSCs (hiPSC-NSPCs) have shown benefits for regenerative therapy of the central
Tijmen H Booij et al.
SLAS discovery : advancing life sciences R & D, 22(8), 974-984 (2017-06-24)
Polycystic kidney disease (PKD) is a prevalent disorder characterized by renal cysts that lead to kidney failure. Various signaling pathways have been targeted to stop disease progression, but most interventions still focus on alleviating PKD-associated symptoms. The mechanistic complexity of
Mariama Jaiteh et al.
Journal of medicinal chemistry, 61(12), 5269-5278 (2018-05-25)
Modulation of multiple biological targets with a single drug can lead to synergistic therapeutic effects and has been demonstrated to be essential for efficient treatment of CNS disorders. However, rational design of compounds that interact with several targets is very
Anish A Kanhai et al.
Scientific reports, 10(1), 1672-1672 (2020-02-06)
Autosomal Dominant Polycystic Kidney Disease (ADPKD) is one of the most common monogenic disorders, characterized by the progressive formation of fluid-filled cysts. Tolvaptan is an approved drug for ADPKD patients, but is also associated with multiple side effects. The peroxisome

Articles

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service