Skip to Content
Merck

Skip To

B7166

5-Bromouridine 5′-triphosphate sodium salt

≥93%, synthetic (organic), powder

Synonym(s):

5-BrUTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H14BrN2O15P3 · xNa+
CAS Number:
Molecular Weight:
563.04 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Assay:
≥93%
Biological source:
synthetic (organic)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Bromouridine 5′-triphosphate sodium salt, ≥93%

biological source

synthetic (organic)

Quality Level

assay

≥93%

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OP(OP(OP(O)(O)=O)(O)=O)(OC[C@H]1O[C@@H](N(C=C2Br)C(NC2=O)=O)[C@H](O)[C@@H]1O)=O.[Na]

InChI

1S/3Na/q3*+1

InChI key

BVPYARCRLTXGBD-UHFFFAOYSA-N

General description

5-Bromouridine 5′-triphosphate (5-BrUTP) is used to measure transcription via labeling of ribonucleic acids (RNA). RNA or cells labeled via 5-BrUTP incorporation may be detected immunologically with antibodies.[1]

Application

5-Bromouridine 5′-triphosphate (5-BrUTP) is used to measure transcription via labeling of ribonucleic acids (RNA). RNA or cells labeled via 5-BrUTP incorporation may be detected immunologically with antibodies.
5-Bromouridine 5′-triphosphate sodium salt has been used:
  • to infer the chromosome intermingling between embryonic stem (ES) cells (differentiated and undifferentiated) and terminally differentiated NIH3T3 cells[2]
  • to label nascent RNA in in vivo transcription analysis using fibroblasts[3]
  • and transfected MDA-MB-231 cells[4]
  • to label RNA for nuclear run-on experiments[5]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B0631D4001M6133
biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

natural (inorganic)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥93%

assay

≥90%

assay

≥95%

assay

≥85% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

-

solubility

water: 50 mg/mL, clear, colorless to faint yellow or tan


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Bianca S Heinrich et al.
PLoS pathogens, 6(6), e1000958-e1000958 (2010-06-30)
Positive-strand and double-strand RNA viruses typically compartmentalize their replication machinery in infected cells. This is thought to shield viral RNA from detection by innate immune sensors and favor RNA synthesis. The picture for the non-segmented negative-strand (NNS) RNA viruses, however
Hong-Thuy Bui et al.
Reproduction (Cambridge, England), 141(1), 67-77 (2010-10-27)
Several lines of evidence indicate that the formation of a transcriptionally repressive state during the two-cell stage in the preimplantation mouse embryo is superimposed on the activation of the embryonic genome. However, it is difficult to determine the profile of
Claire Tolza et al.
Molecular cancer research : MCR, 17(10), 1999-2014 (2019-07-14)
The architectural chromatin protein HMGA1 and the transcription factor Fra-1 are both overexpressed in aggressive triple-negative breast cancers (TNBC), where they both favor epithelial-to-mesenchymal transition, invasion, and metastasis. We therefore explored the possibility that Fra-1 might be involved in enhanced



Global Trade Item Number

SKUGTIN
B7166-50MG04061833002155
B7166-5MG04061833441596
B7166-10MG04061833002148

Questions

Reviews

No rating value

Active Filters