Skip to Content
Merck
All Photos(1)

Key Documents

M6133

Sigma-Aldrich

7-Methylguanosine 5′-triphosphate sodium salt

≥85% (HPLC)

Synonym(s):

m7GTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N5O14P3 · xNa+
CAS Number:
Molecular Weight:
537.21 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

natural (inorganic)

Assay

≥85% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na+].C[n+]1cn([C@@H]2O[C@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O)c3N=C(N)NC([O-])c13

InChI

1S/C11H20N5O14P3.Na/c1-15-3-16(8-5(15)9(19)14-11(12)13-8)10-7(18)6(17)4(28-10)2-27-32(23,24)30-33(25,26)29-31(20,21)22;/h3-4,6-7,9-10,17-18H,2H2,1H3,(H,23,24)(H,25,26)(H3,12,13,14)(H2,20,21,22);/q;+1/p-1/t4-,6-,7-,9?,10-;/m1./s1

InChI key

FSMRTZAMTMQMEF-OCMBMSOASA-M

General description

7-Methylguanosine 5′-triphosphate is a mRNA cap structural analog.

Application

7-Methylguanosine 5′-triphosphate (m7GTP) is use to study the structures and mechanisms of eukaryotic cellular mRNA caps in the context of mRNA involvement in protein synthesis. m7GTP may be used to create affinity media for capture and purification of molecules such as eukaryotic initiation factor eIF4E.
7-Methylguanosine 5′-triphosphate sodium salt has been used to test its affinity towards Poly(A)-specific ribonuclease (PARN) polypeptides using fluorescence titrations. It has also been used in crystallization along with gemin5 fragment (G5N) and DM15 region of human La-related 15 protein 1 ( LARP1).

Biochem/physiol Actions

7-Methylguanosine 5′-triphosphate (m7GTP) inhibits in vitro protein synthesis by binding to translation initiation complexes. m7GTP is useful to study the structures and mechanisms of eukaryotic cellular mRNA caps in the context of mRNA involvement in protein synthesis. m7GTP has a high affinity for eukaryotic initiation factor eIF4E and may be used to create affinity media for their capture and purification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Jun Wang et al.
PloS one, 7(9), e44079-e44079 (2012-09-08)
Influenza virus, which spreads around the world in seasonal epidemics and leads to large numbers of deaths every year, has several ribonucleoproteins in the central core of the viral particle. These viral ribonucleoproteins can specifically bind the conserved 3' and
Yih-Leh Huang et al.
The Journal of biological chemistry, 280(13), 13153-13162 (2005-01-29)
The S-adenosylmethionine-dependent guanylyltransferase of bamboo mosaic virus belongs to a novel class of mRNA capping enzymes distantly conserved in Alphavirus-like superfamily. The reaction sequence of the viral enzyme has been proposed comprising steps of 1) binding of GTP and S-adenosylmethionine
Regina Cencic et al.
Analytical biochemistry, 560, 24-29 (2018-09-09)
All cellular cytoplasmic mRNAs are capped at their 5' ends with an m7GpppN group. Several proteins that mediate cap function have been identified by cap affinity purification, enabling their characterization in a number of biological processes. Among these, eukaryotic initiation
The DEAD-box helicase DDX3 substitutes for the cap-binding protein eIF4E to promote compartmentalized translation initiation of the HIV-1 genomic RNA
Soto-Rifo R, et al.
Nucleic Acids Research, 41(12), 6286-6299 (2013)
Alan L Ho et al.
PloS one, 7(7), e40439-e40439 (2012-07-19)
Uveal melanomas possess activation of the mitogen-activated protein kinase (MAPK) and phosphoinositide 3-kinase (PI3K)/AKT/mammalian Target of Rapamycin (mTOR) pathways. MAPK activation occurs via somatic mutations in the heterotrimeric G protein subunits GNAQ and GNA11 for over 70% of tumors and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service