Skip to Content
Merck
All Photos(3)

Key Documents

D4001

Sigma-Aldrich

2′-Deoxyuridine 5′-triphosphate sodium salt

≥95%

Synonym(s):

dUTP sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N2O14P3 · xNa+
CAS Number:
Molecular Weight:
468.14 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O14P3.Na.H/c12-5-3-8(11-2-1-7(13)10-9(11)14)23-6(5)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,12H,3-4H2,(H,18,19)(H,20,21)(H,10,13,14)(H2,15,16,17);;

InChI key

FVFWWPFKJNTHIQ-UHFFFAOYSA-N

Application

2′-Deoxyuridine 5′-triphosphate (dUTP) may be used to study the distribution, specificity and kinetics of deoxyuridine 5′-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.dUTP is a component of the TUNEL (terminal deoxyribonucleotidyl-transferase-(TdT)-mediated deoxyuridine-5′-triphosphate- (dUTP) nick-end labeling) test. TUNEL is a common method for detecting DNA fragmentation that results from apoptotic signaling cascades.
2′-Deoxyuridine 5′-triphosphate sodium salt has been used in the extraction of white muscles from the African catfish.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Claudia Ulbrich et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 26(6), 1011-1022 (2011-01-12)
Myocardium is very sensitive to gravitational changes. During a spaceflight cardiovascular atrophy paired with rhythm problems and orthostatic intolerance can occur. The aim of this study was to investigate the impact of basic fibroblast growth factor (bFGF) and vascular endothelial
Kwou-Yeung Wu et al.
Molecular vision, 14, 1705-1712 (2008-09-23)
Previous studies have indicated that improper use of mitomycin C (MMC) caused cytotoxicity in corneal endothelial cells. The aim of the present study was to investigate whether MMC induces cellular apoptosis in corneal endothelial cells and to determine the mechanism
Alexandra Kalogeraki et al.
Acta cytologica, 54(1), 39-42 (2010-03-24)
To evaluate apoptosis and cell proliferation on cytologic specimens (smears) from endoscopic retrograde cholangiopancreatography in patients with ampulary carcinoma and to correlate that relationship with the grade of the tumors. Forty patients (23 males and 17 females) aged 45-81 who
Dietary canitine maintains energy reserves and delays fatigue of exercised african catfish (Clarias gariepinus) fed high fat diets
Ozorio R, et al.
Scientia Agricola, 62(3), 208-213 (2005)
Indalecio Quesada-Soriano et al.
Biochimie, 92(2), 178-186 (2009-11-03)
Deoxyuridine 5'-triphosphate nucleotidohydrolase (dUTPase, EC 3.6.1.23) catalyzes the hydrolysis of dUTP to dUMP and pyrophosphate, and plays important roles in nucleotide metabolism and DNA replication. Hydrolysis of other nucleotides similar in structure to dUTP would be physiologically negative and therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service