Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
3
Pricing and availability is not currently available.
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human, rat, mouse
technique(s)
indirect immunofluorescence: 10 μg/mL using CV-1 cells, western blot: 2 μg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... VAMP4(8674)
mouse ... Vamp4(53330)
rat ... Vamp4(364033)
General description
Vesicle associated membrane protein 4 (VAMP4) is localized on human chromosome 1q24-q25. It is expressed in the skeletal muscle and liver. The protein is part of the soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNARE) family. VAMP4 is localized to the trans-Golgi network.
Immunogen
recombinant rat VAMP-4.
Biochem/physiol Actions
Anti-Vesicle Associated Membrane Protein 4 detectsVAMP-4.
Vesicle associated membrane protein 4 (VAMP4) mediates trans-Golgi network trafficking of membrane-bound and secreted proteins. Studies have shown that VAMP4 has a role in maintaining the structure of the Golgi apparatus.
Physical form
Solution in phosphate buffered saline containing 1.0 mg/mL bovine serum albumin and 0.05% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.