Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
(CH3)3SiNHSi(CH3)3
CAS Number:
Molecular Weight:
161.39
EC Number:
213-668-5
UNSPSC Code:
12352300
PubChem Substance ID:
Beilstein/REAXYS Number:
635752
MDL number:
Pricing and availability is not currently available.
assay
≥97% (GC)
form
liquid
refractive index
n20/D 1.407 (lit.)
bp
125 °C (lit.)
storage temp.
2-8°C
SMILES string
C[Si](C)(C)N[Si](C)(C)C
InChI
1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3
InChI key
FFUAGWLWBBFQJT-UHFFFAOYSA-N
Application
A reagent for the preparation of trimethylsilyl derivatives.
Still not finding the right product?
Explore all of our products under Hexamethyldisilazane
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
52.5 °F - closed cup
flash_point_c
11.4 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ashang Luwang Laiva et al.
International journal of pharmaceutics, 483(1-2), 115-123 (2015-02-15)
Electrospun hybrid nanofibrous scaffolds have gained much importance in the field of tissue engineering and drug delivery applications owing to its multifaceted properties. In this study, the properties of composite polycaprolactone (PCL)/silk fibroin (SF) nanofibrous scaffolds was investigated as a
Niklas Söderholm et al.
PloS one, 6(9), e24353-e24353 (2011-09-29)
Polymorphonuclear neutrophils (PMNs) are important components of the human innate immune system and are rapidly recruited at the site of bacterial infection. Despite the effective phagocytic activity of PMNs, Neisseria gonorrhoeae infections are characterized by high survival within PMNs. We
Claire Yu et al.
Biomaterials, 194, 1-13 (2018-12-19)
Decellularized extracellular matrices (dECMs) have demonstrated excellent utility as bioscaffolds in recapitulating the complex biochemical microenvironment, however, their use as bioinks in 3D bioprinting to generate functional biomimetic tissues has been limited by their printability and lack of tunable physical

