Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C17H27NO3
CAS Number:
Molecular Weight:
293.40
EC Number:
219-484-1
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
2144300
MDL number:
Pricing and availability is not currently available.
storage temp.
2-8°C
SMILES string
CCCCCCCCC(=O)NCc1ccc(O)c(OC)c1
InChI
1S/C17H27NO3/c1-3-4-5-6-7-8-9-17(20)18-13-14-10-11-15(19)16(12-14)21-2/h10-12,19H,3-9,13H2,1-2H3,(H,18,20)
InChI key
RGOVYLWUIBMPGK-UHFFFAOYSA-N
Gene Information
rat ... Trpv1(83810), Trpv4(66026)
Still not finding the right product?
Explore all of our products under N-Vanillylnonanamide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the


