Skip to Content
Merck

Skip To

A6631

Potassium arsenate monobasic

Synonym(s):

Arsenic acid potassium salt, Potassium dihydrogen arsenate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
KH2AsO4
CAS Number:
Molecular Weight:
180.03
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
232-065-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

reaction suitability

reagent type: catalyst
core: arsenic

mp

277-283 °C (lit.)

SMILES string

[K+].O[As](O)([O-])=O

InChI

1S/AsH3O4.K/c2-1(3,4)5;/h(H3,2,3,4,5);/q;+1/p-1

InChI key

GVPLVOGUVQAPNJ-UHFFFAOYSA-M

Application

Potassium arsenate may be used as a reference material during the analysis of biological materials for the presence of arsenate. Potassium arsenate is used as a metabolic inhibitor and toxicity agent.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S Gautami et al.
Journal of toxicology. Clinical toxicology, 33(2), 131-133 (1995-01-01)
An accidental food poisoning outbreak occurred resulting in the death of 14 of the 22 affected persons. Chemical analysis of food materials revealed the presence of sodium nitrite and potassium arsenate. Analysis of urine, gastric contents, liver and kidney, collected
Jian J Duan et al.
Environmental entomology, 36(4), 982-988 (2007-08-25)
A 13-d continuous dietary exposure bioassay using nymphs of the insidious flower bug, Orius insidiosus (Say) (Heteroptera: Anthocoridae), was developed to assess the potential dietary effects of insecticidal substances that have little or no contact toxicity. The nymphs were fed
Jianguo Tan et al.
Environmental entomology, 40(6), 1613-1621 (2012-01-06)
The insidious flower bug, Orius insidiosus (Say) (Heteroptera: Anthocoridae) is an important surrogate species for assessing potential effects of plant-incorporated protectants (PIPs) on nontarget heterotrophic predators. In this study, a continuous dietary exposure system was optimized by assessing the effect



Global Trade Item Number

SKUGTIN
A6631-5G04061833382585
A6631-100G04061833382561
A6631-50G04061833382578

Questions

Reviews

No rating value

Active Filters