Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.56
UNSPSC Code:
13111000
Form:
suspension
Pricing and availability is not currently available.
form
suspension
technique(s)
affinity chromatography: suitable
mean particle size
70 μm
storage temp.
2-8°C
Application
Byzen Pro™ Protein A resin is an affinity chromatography medium in which a novel Protein A mutant is used as a ligand. Conventional protein A resins use strongly acidic buffers to elute antibodies, exposing the target antibody to the risks of aggregation and decreased activities. In contrast, this temperature- responding Byzen Pro™ Protein A resin allows antibodies to be eluted under neutral pH by a simple temperature change. This eliminates any risk of aggregation and decreased activity of the antibody, improving manufacturing efficiency for pharmaceutical antibodies.
Features and Benefits
Typical Product Profile:
Bead Structure, Cross-linked polyvinyl alcohol, Mean Particle Size 70 μm, Max. Back Pressure 0.3 MPa, Flow Rate (Recommended) 125-250 cm/h, pH Range (Recommended) pH 6-8, Ligand: Recombinant Protein A mutant, Ligand Immobilization: Multiple ligation, Dynamic Binding Capacity (20 cm column): 26 g/L (250 cm/h) 34 g/L (125 cm/h), Total Binding Capacity 40 g/L, Binding Temperature Below 10°C, Eluting Temperature 37 to 40°C, Mode of Temperature Change: Conventional heat exchange system can be used.
Bead Structure, Cross-linked polyvinyl alcohol, Mean Particle Size 70 μm, Max. Back Pressure 0.3 MPa, Flow Rate (Recommended) 125-250 cm/h, pH Range (Recommended) pH 6-8, Ligand: Recombinant Protein A mutant, Ligand Immobilization: Multiple ligation, Dynamic Binding Capacity (20 cm column): 26 g/L (250 cm/h) 34 g/L (125 cm/h), Total Binding Capacity 40 g/L, Binding Temperature Below 10°C, Eluting Temperature 37 to 40°C, Mode of Temperature Change: Conventional heat exchange system can be used.
Legal Information
Byzen Pro is a trademark of Nomadic Bioscience Co., Ltd
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) affinity chromatography: suitable | technique(s) affinity chromatography: suitable | technique(s) - | technique(s) affinity chromatography: suitable |
| form suspension | form saline suspension | form aqueous suspension | form lyophilized powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| mean particle size 70 μm | mean particle size - | mean particle size - | mean particle size - |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Manual for Antibody Purification by Affinity Chromatography using 18887 Byzen ProTM Protein A resin
Ichiro Koguma et al.
Journal of chromatography. A, 1305, 149-153 (2013-07-31)
We attempted to evaluate a novel purification method of immunoglobulins (IgGs) by using a mutant type of protein A. Although this mutant protein A binds to IgGs at 5°C, the IgGs are released at 40°C; hence, it was designated as
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 532134-25G | 04061831820812 |
| 18887-5ML | 04061838212191 |
| 18887-1ML | 04061838089939 |
| 18887-25ML | 04061838083944 |
| 18887-2ML | 04061836971038 |



