Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
product line
BioReagent
form
solid
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 625 nm; λem 646 nm
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto Rho14 maleimide is a new rhodamine featuring a functionality for coupling to biomolecules such as DNA, RNA or proteins. The label shows strong absorption, and extraordinarily high fluorescence quantum yield. In fact Atto Rho14 is the brightest label available in this wavelength range. In addition the dye exhibits an exceptionally high photostability.
After coupling to a substrate Atto Rho14 maleimide carries a net electrical charge of +1.
Absorption and fluorescence are pH-independent in the range of pH 2 to 11, used in typical applications. The dye is moderately hydrophilic.
find more information here
After coupling to a substrate Atto Rho14 maleimide carries a net electrical charge of +1.
Absorption and fluorescence are pH-independent in the range of pH 2 to 11, used in typical applications. The dye is moderately hydrophilic.
find more information here
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Storage Class
13 - Non Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 538817-5G | 04061832287737 |