Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
(CH3)2C(CH2OH)N(CH3)2
CAS Number:
Molecular Weight:
117.19
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1732837
Pricing and availability is not currently available.
grade
technical
assay
~80% (NT)
impurities
~20% water
refractive index
n20/D 1.447
bp
158-160 °C (lit.)
density
0.95 g/mL at 20 °C (lit.)
SMILES string
CN(C)C(C)(C)CO
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.