Skip to Content
Merck

Skip To

AB2277

Anti-FOXP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

fork head-related protein like B, forkhead box P1, glutamine-rich factor 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rhesus macaque, rat, mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXP1(27086)

General description

FOXP1 has 4 isoforms observed at 75/73, 64 and 54 kDa
Forkhead box protein 1 (FOXP1) is a member of the Forkhead family of transcription factors, which have been characterized as transcriptional repressors and activators, and as having a DNA-binding domain distinguished by its formation of a winged-helix motif. FOXP1 is known to be an important regulator of airway morphogenesis, lung epithelial development, and early cardiac development. It is also known to have an important role in conjunction with NFκβ in the development of B-cells and the differentiation of macrophages. Splice variants give rise to four isoforms of FOXP1. The protein is known to form homodimers and heterodimers with FOXP2 and FOXP4. Studies have indicated FOXP1 as a potential inducer of oncogenesis when over expressed. The depletion and mutation of FOXP1 expression may lead to defects in embryonic cardiac, lung and esophagus epithelial development.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human FOXP1.

Application

Anti-FOXP1 Antibody detects level of FOXP1 & has been published & validated for use in WB, IC, IH.
Immunohistochemistry Analysis: A 1:300 dilution of AB2277 from a previous lot detected FOXP1 in inflammatory skin disorder and normal human kidney-glomerulus tissue.

Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected FOXP1 in NIH/3T3, RAW264.7, and HeLa cells.
Research Category
Neuroscience

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Transcription Factors

Biochem/physiol Actions

This antibody recognizes the C-terminus of FOXP1.

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.
Affinity purified

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected multiple isoforms of FOXP1 in 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV32564SAB5500108WH0027086M1
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

flow cytometry: 1:100, immunoblotting: 1:25, immunohistochemistry: 1:100

technique(s)

indirect ELISA: suitable, western blot: 1-5 μg/mL

UniProt accession no.

Q9H334

UniProt accession no.

Q9H334

UniProt accession no.

Q9H334

UniProt accession no.

Q9H334


Still not finding the right product?

Explore all of our products under Anti-FOXP1 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters