Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
goat
Quality Level
conjugate
peroxidase conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
shipped in
dry ice
target post-translational modification
unmodified
Application
Secondary & Control Antibodies
Whole Immunoglobulin Secondary Antibodies
Physical form
Preparation Note
Analysis Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity purified immunoglobulin | antibody form - | antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form - |
| conjugate peroxidase conjugate | conjugate - | conjugate peroxidase conjugate | conjugate peroxidase conjugate |
| species reactivity mouse | species reactivity - | species reactivity pig | species reactivity mouse |
| technique(s) western blot: suitable | technique(s) - | technique(s) ELISA: suitable, western blot: suitable | technique(s) ELISA: suitable, western blot: suitable |
| biological source goat | biological source - | biological source goat | biological source goat |
| shipped in dry ice | shipped in - | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



